Ion AmpliSeq™ Transcriptome Mouse Gene Expression Panel, Chef-Ready Kit
Ion AmpliSeq™ Transcriptome Mouse Gene Expression Panel, Chef-Ready Kit
Ion Torrent™
Ion AmpliSeq™ Transcriptome Mouse Gene Expression Panel, Chef-Ready Kit
The Ion AmpliSeq Transcriptome Mouse Gene Expression Panel, Chef-Ready Kit allows simultaneous gene expression measurement of over 20,000 mouse RefSeqRead more
Have Questions?
Catalog NumberQuantity
A3641232 reactions
Catalog number A36412
Price (USD)
7,940.00
Each
Add to cart
Quantity:
32 reactions
Price (USD)
7,940.00
Each
Add to cart
The Ion AmpliSeq Transcriptome Mouse Gene Expression Panel, Chef-Ready Kit allows simultaneous gene expression measurement of over 20,000 mouse RefSeq genes in a single assay. Providing the ability to work directly with low input levels of total RNA, the Ion AmpliSeq Transcriptome Mouse Gene Expression Assay is ideal for those interested in obtaining comprehensive and highly sensitive gene expression measurements from a wide range of RNA sources. This particular kit enables automated production of Ion AmpliSeq transcriptome libraries on the Ion Chef System. For manual production, see the Ion AmpliSeq Transcriptome Mouse Gene Expression Kit.
The Ion AmpliSeq Transcriptome Mouse Gene Expression Panel, Chef-Ready Kit offers:
• Flexible, straightforward library preparation—start with as little as 10 ng of total RNA from tissues, cells, or other sources
• Fast gene expression profiling with simple workflow—go from RNA to data in <2 days with ∼45 minutes of total hands-on time with the automated workflow of the Ion GeneStudio S5 and Ion Chef systems
• A complementary software plug-in for automated sequencing analysis that provides gene quantitation and reporting as well as quality control metrics
• Compatibility with Ion Xpress Barcode adapters, allowing for multiplexed sequencing
Leveraging the speed and simplicity of Ion AmpliSeq technology, this ultrahigh-multiplex PCR enrichment approach enables rapid gene expression profiling with minimal hands-on time. This research method provides a faster, simpler, and lower-cost sequencing method that is highly correlated to whole-transcriptome RNA sequencing.
Straightforward library preparation and multiplex sequencing with Chef-Ready Kit
The Ion AmpliSeq Transcriptome Mouse Gene Expression Panel, Chef-Ready Kit, includes the reagents and materials required for automated preparation of 32 barcoded Ion AmpliSeq transcriptome libraries. The Ion Chef System can automate up to eight barcoded libraries per run, so the kit provides reagents and materials sufficient for four runs. Starting with as little as 10 ng of total RNA, the total RNA is first processed using the SuperScript VILO cDNA Synthesis Kit (not included), and the resulting cDNA amplified using Ion AmpliSeq technology to accurately maintain expression levels of all genes. Approximately 7 hours of instrument run time and only 15 minutes of hands-on time are needed to set up the Ion Chef System for library construction from cDNA.
The resulting libraries are ready for downstream template preparation using the Ion Chef System and sequencing on the Ion GeneStudio S5 or Ion Proton sequencing system. With the recommended 16 libraries per Ion 550 Chip and 8 libraries per Ion 540 or Ion PI chip, libraries are multiplexed for flexibility, throughput increase, and cost reduction. Other multiplexing options are also possible, depending on the experimental design and desired level of sensitivity.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
LibrariesTargeted Sequencing Library
Product TypeTranscriptome Mouse Gene Expression Panel Chef-Ready Kit
Quantity32 reactions
Shipping ConditionDry Ice
Workflow StepLibrary Generation
Product LineIon AmpliSeq™
Starting MaterialRNA
Unit SizeEach
Contents & Storage
• Ion AmpliSeq Transcriptome Mouse Gene Expression Panel for Chef-Ready Kit
• Ion AmpliSeq Kit for Chef DL8
Store in freezer (-5 to -30°C).
Frequently asked questions (FAQs)
How many reactions can I perform with the Ion AmpliSeq RNA ERCC Companion Panel?
The Ion AmpliSeq RNA ERCC Companion Panel provides enough reagents to perform 96 manual reactions or 48 automated reactions.
What software do I need to view data from the Ion AmpliSeq RNA ERCC Companion Panel?
To view data from the Ion AmpliSeq RNA ERCC Companion Panel, Torrent Suite Software version 5.10 or later is required and Version 5.10.0.3 or later of the ERCC_Analysis plugin for Torrent Suite Software is required.
Can the Ion AmpliSeq RNA ERCC Companion Panel be used with FFPE samples?
We do not recommend using the Ion AmpliSeq RNA ERCC Companion Panel with FFPE samples or with any panels besides human and mouse Ion AmpliSeq transcriptome gene expression panels.
How much of the Ion Ampliseq RNA ERCC Companion Panel should I add to the target amplification reactions?
We recommend adding 1 µL of the Ion Ampliseq RNA ERCC Companion Panel per manual target amplification reaction and 8 µL per primer pool for an automated target amplification reaction on the Ion Chef Instrument.
Why would one use the Ion AmpliSeq ERCC Companion Panel?
The Ion AmpliSeq ERCC Companion Panel is compatible with Ion AmpliSeq Transcriptome Human Gene Expression Kit and Ion AmpliSeq Transcriptome Mouse Gene Expression Kit (not recommended for use with FFPE samples or panels outside of Ion AmpliSeq Human or Mouse Transcriptome kits). With the addition of the Ion AmpliSeq RNA ERCC Companion panel to your Ion AmpliSeq RNA panel, dynamic range and sensitivity can be similarly evaluated in Ion AmpliSeq RNA libraries that are prepared from RNA samples containing the ERCC RNA Spike-In Mix
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link