Ion AmpliSeq™ Transcriptome Mouse Gene Expression Kit
Ion AmpliSeq™ Transcriptome Mouse Gene Expression Kit
Ion Torrent™

Ion AmpliSeq™ Transcriptome Mouse Gene Expression Kit

The Ion AmpliSeq Transcriptome Mouse Gene Expression Assay allows simultaneous gene expression measurement of over 20,000 mouse RefSeq genes inRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
A3655496 Reactions
A3655324 Reactions
A36555384 Reactions
Catalog number A36554
Price (USD)
16,020.00
Each
Add to cart
No. of Reactions:
96 Reactions
Price (USD)
16,020.00
Each
Add to cart
The Ion AmpliSeq Transcriptome Mouse Gene Expression Assay allows simultaneous gene expression measurement of over 20,000 mouse RefSeq genes in a single assay. Providing the ability to work directly with low input levels of total RNA, the Ion AmpliSeq Transcriptome Mouse Gene Expression Assay is ideal for those interested in obtaining comprehensive and highly sensitive gene expression measurements from a wide range of RNA sources. This particular kit enables manual library preparation with throughput flexibility. For automated production, see the Ion AmpliSeq Transcriptome Mouse Gene Expression Panel, Chef-Ready Kit.

The Ion AmpliSeq Transcriptome Mouse Gene Expression Assay offers:
• Flexible, straightforward library preparation—start with as little as 10 ng of total RNA from tissues, cells, or other sources
• Fast gene expression profiling with simple workflow—go from RNA to data in <2 days with ∼45 minutes of total hands-on time with the automated workflow of the Ion GeneStudio S5 and Ion Chef systems
• A complementary software plug-in for automated sequencing analysis that provides gene quantitation and reporting as well as quality control metrics
• Compatibility with Ion Xpress Barcode adapters, allowing for multiplexed sequencing

Leveraging the speed and simplicity of Ion AmpliSeq technology, this ultrahigh-multiplex PCR enrichment approach enables rapid gene expression profiling with minimal hands-on time. This research method provides a faster, simpler, and lower-cost sequencing method that is highly correlated to whole-transcriptome RNA sequencing.

Straightforward library preparation and multiplex sequencing with manual kit
Starting with as little as 10 ng of total RNA, libraries can be prepared in approximately seven hours with only one hour of hands-on time. Total RNA is first processed with the SuperScript VILO cDNA Synthesis Kit (not included), and the resulting cDNA is amplified using Ion AmpliSeq technology to accurately maintain expression levels of all genes.

The resulting libraries are ready for downstream template preparation using the Ion OneTouch 2 or Ion Chef System and sequencing on the Ion GeneStudio S5 or Ion Proton System. Libraries can be barcoded for multiplexing, providing flexibility, reduced cost, and increased throughput, with the recommended 16 libraries per Ion 550 Chip and 8 libraries per Ion 540 or Ion PI chip. Other multiplexing options are also possible, depending on the experimental design and desired level of sensitivity.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormatKit
LibrariesTargeted Sequencing Library
Product LineIon AmpliSeq™
Product TypeTranscriptome Mouse Gene Expression Kit
Quantity96 reactions
Shipping ConditionDry Ice
SpeciesMouse
For Use With (Application)Gene Expression, miRNA analysis
No. of Reactions96 Reactions
Sample TypeRNA
Unit SizeEach
Contents & Storage
Store in freezer at -5 to -30°C.

Frequently asked questions (FAQs)

How many reactions can I perform with the Ion AmpliSeq RNA ERCC Companion Panel?

The Ion AmpliSeq RNA ERCC Companion Panel provides enough reagents to perform 96 manual reactions or 48 automated reactions.

What software do I need to view data from the Ion AmpliSeq RNA ERCC Companion Panel?

To view data from the Ion AmpliSeq RNA ERCC Companion Panel, Torrent Suite Software version 5.10 or later is required and Version 5.10.0.3 or later of the ERCC_Analysis plugin for Torrent Suite Software is required.

Can the Ion AmpliSeq RNA ERCC Companion Panel be used with FFPE samples?

We do not recommend using the Ion AmpliSeq RNA ERCC Companion Panel with FFPE samples or with any panels besides human and mouse Ion AmpliSeq transcriptome gene expression panels.

How much of the Ion Ampliseq RNA ERCC Companion Panel should I add to the target amplification reactions?

We recommend adding 1 µL of the Ion Ampliseq RNA ERCC Companion Panel per manual target amplification reaction and 8 µL per primer pool for an automated target amplification reaction on the Ion Chef Instrument.

Why would one use the Ion AmpliSeq ERCC Companion Panel?

The Ion AmpliSeq ERCC Companion Panel is compatible with Ion AmpliSeq Transcriptome Human Gene Expression Kit and Ion AmpliSeq Transcriptome Mouse Gene Expression Kit (not recommended for use with FFPE samples or panels outside of Ion AmpliSeq Human or Mouse Transcriptome kits). With the addition of the Ion AmpliSeq RNA ERCC Companion panel to your Ion AmpliSeq RNA panel, dynamic range and sensitivity can be similarly evaluated in Ion AmpliSeq RNA libraries that are prepared from RNA samples containing the ERCC RNA Spike-In Mix

Share catalog number, name or link