Informational:Inquire about OEM or Commercial Supply version of this product here.
Applied Biosystems™

MagMAX™ DNA Multi-Sample Ultra 2.0 Binding Beads

Catalog number: A36579
Have Questions?
Applied Biosystems™

MagMAX™ DNA Multi-Sample Ultra 2.0 Binding Beads

Catalog number: A36579
Have Questions?
Catalog Number
A36579
Unit Size
45 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A3657945 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
These Magnetic Binding beads are included in the MagMAX DNA Multi-Sample Ultra 2.0 Kit. They are available separately for applications that require more binding beads than are provided in the kit.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Line
MagMAX™
Product Type
DNA Multi-Sample Ultra 2.0 Binding Bead
Quantity
45 mL
Shipping Condition
Room Temperature

Contents & Storage

Store at room temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text