Informational:Inquire about OEM or Commercial Supply version of this product here.
Applied Biosystems™

MagMAX™ DNA Multi-Sample Ultra 2.0 Lysis/Binding Solution

Catalog number: A36581
Have Questions?
Applied Biosystems™

MagMAX™ DNA Multi-Sample Ultra 2.0 Lysis/Binding Solution

Catalog number: A36581
Have Questions?
Catalog Number
A36581
Unit Size
450 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A36581450 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
This Lysis/Binding solution is included in the MagMAX DNA Multi-Sample Ultra 2.0 Kit. It is available separately for applications that require more Lysis/Binding solution than is provided in the kit.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Line
MagMAX™
Product Type
DNA Multi-Sample Ultra 2.0 Lysis/Binding Solution
Quantity
450 mL
Shipping Condition
Room Temperature

Contents & Storage

Store at room temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text