Thermo Scientific™

NeuCode™ Lysine-521

Catalog number: A36753
Have Questions?
Thermo Scientific™

NeuCode™ Lysine-521

Catalog number: A36753
Have Questions?
Catalog Number
A36753
Unit Size
25 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A3675325 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific NeuCode amino acids augment the level of multiplexing achievable for the metabolic labeling of proteins for mass spectrometry analysis. NeuCode Lysine-521 (1,2,3,4,5-13C5, 6,6-D2,15N L-Lysine-2HCl) may also be used with traditional SILAC to improve flexibility of multiplexing options or to reduce complexity of analysis.

General features of NeuCode SILAC labeling:
• Labeling efficiency—100% label incorporation into proteins of living cells without toxicity
• Compatible—may be multiplexed with existing SILAC amino acids
• Time-saving—not necessary to label to 100% incorporation if only using heavy amino acids
• High-quality supplements—heavy amino acids with >98% isotope purity

Stable isotope labeling with amino acids in cell culture (SILAC) is a powerful method to identify and quantify relative differential changes in complex protein samples. NeuCode metabolic labeling is similar to SILAC, but differs in that the labeling only utilizes heavy amino acids. The increased multiplexing capability of NeuCode amino acids is possible through the use of mass defects from extra neutrons in the stable isotopes. These small mass differences may be resolved on high resolution mass spectrometers (Thermo Scientific™ Orbitrap™ Elite™, Orbitrap™ Fusion™ Tribrid™, and Orbitrap™ Fusion™ Lumos™ Tribrid™ mass spectrometers). Use of only heavy amino acids eliminates the need for 100% incorporation of amino acids used for SILAC (both heavy and light), and may be especially useful for studies with primary cells.

NeuCode amino acids are used together with specialized cell culture media that are deficient in essential amino acids. Heavy L-lysine is used for SILAC analysis of peptides that have been digested with trypsin or LysC. NeuCode Lysine-521 may be used with 13C6 15N2 L-Lysine-2HCl (NeuCode Lysine-602), NeuCode Lysine-440, and NeuCode Lysine-080 for NeuCode four-plex experiments and may be combined with light lysine and 4,4,5,5-D4 L-Lysine for three-plex experiments.

Related products
L-Lysine-2HCl, 4,4,5,5-D4
13C6 15N2 L-Lysine-2HCl
NeuCode Lysine-080
Neucode Lysine-440

NeuCode is a trademark of WARF.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Final Product Type
Protein
Product Line
NeuCode™
Product Type
Lysine
Quantity
25 mg
Format
Solid
Label or Dye
Mass Isotope of Amino Acid
Shipping Condition
Room Temperature
Form
Solid

Contents & Storage

Store at room temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text