Pierce™ 3x DYKDDDDK Peptide
Pierce™ 3x DYKDDDDK Peptide
Thermo Scientific™

Pierce™ 3x DYKDDDDK Peptide

Pierce 3X DYKDDDDK Peptide is a lyophilized peptide with the sequence Met-Asp-Tyr-Lys-Asp-His-Asp-Gly-Asp-Tyr-Lys-Asp-His-Asp-Ile-Asp-Tyr-Lys-Asp-Asp-Asp-Asp-Lys (MDYKDHDGDYKDHDIDYKDDDDK). It has a molecular weight of 2861.9Read more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A368065 x 5 mg
A368055 mg
Catalog number A36806
Price (USD)
1,212.65
Online Exclusive
1,364.00
Save 151.35 (11%)
Each
-
Add to cart
Quantity:
5 x 5 mg
Price (USD)
1,212.65
Online Exclusive
1,364.00
Save 151.35 (11%)
Each
Add to cart

Pierce 3X DYKDDDDK Peptide is a lyophilized peptide with the sequence Met-Asp-Tyr-Lys-Asp-His-Asp-Gly-Asp-Tyr-Lys-Asp-His-Asp-Ile-Asp-Tyr-Lys-Asp-Asp-Asp-Asp-Lys (MDYKDHDGDYKDHDIDYKDDDDK). It has a molecular weight of 2861.9 g/mol. For use, reconstitute to 1.5 mg/mL in PBS.
 

This tricosa-peptide can be used to competitively elute DYKDDDDK-tagged fusion proteins from immobilized anti-DYKDDDDK affinity supports, especially when a DYKDDDDK-tagged protein is sensitive to low pH. At a 1.5 mg/mL concentration, complete release of tagged protein can be achieved in 5–10 minutes at ambient temperature.
 

Note: Pierce 3X DYKDDDDK Peptide may interfere with protein determination assays and absorbance at 280 nm. However, the peptide can be removed from the eluate by desalting (e.g., with Zeba Spin Desalting Columns, Cat. No. 89891).

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Protein Purification and Isolation
FormLyophilized Powder
FormatLyophilized Powder
Product LinePierce™
Product TypePeptides
Protein FormRecombinant
Quantity5 x 5 mg
Shipping ConditionWet Ice
MatrixAffinity Resin
Unit SizeEach
Contents & Storage
5 x 5 mg lyophilized peptide. Store at -5 to -30°C.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link