Applied Biosystems™
PrepFiler™ Spin Tubes and Filter Columns, ethylene oxide-treated
Catalog number: A36853
Have Questions?
Applied Biosystems™
PrepFiler™ Spin Tubes and Filter Columns, ethylene oxide-treated
Catalog number: A36853
Have Questions?
Catalog Number
A36853
Unit Size
100 columns/300 tubes
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A36853100 columns/300 tubes
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
PrepFiler Spin Tubes and Filter Columns are available here as a separate purchase, providing ordering flexibility and allowing researchers to select only the plastic consumables needed for use with all PrepFiler forensic DNA extraction kits. These kits use magnetic particles with an optimized multi-component surface chemistry to deliver superior DNA yield from most forensic sample types, including body fluids, stains, and swabs of body fluids.
PrepFiler Filter Columns are used for substrate removal, and PrepFiler Spin Tubes are used for sample processing in tube format. The packaged Spin Tubes and Filter Columns are treated with ethylene oxide.
PrepFiler forensic DNA extraction kits contain other components required for DNA recovery from both routine and challenging samples, including removal of PCR inhibitors.
Specifications
Product Line
PrepFiler
Starting Material
DNA
Product Type
Spin Tube and Filter Column
Shipping Condition
Room Temperature
Contents & Storage
Store tubes and columns at ambient temperature.