Ion Torrent™

Ion 550™ Single Chip Supplemental Kit

Catalog number: A36953
Have Questions?
Ion Torrent™

Ion 550™ Single Chip Supplemental Kit

Catalog number: A36953
Have Questions?
Catalog Number
A36953
Unit Size
4 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A369534 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Ion 550 Single Chip Supplemental Kit contains pre-packaged templating supplies and sequencing reagent cartridges required for use with the single-chip Ion Chef workflow. The Ion 550 Single Chip Supplemental Kit, together with the Ion 550 Kit—Chef, enables the flexibility to prepare one loaded chip per Ion Chef run, without loss of templating reagents.

Note: This supplemental templating kit for Ion Chef is only compatible with the Ion 550 Chip and the Ion 550 Kit—Chef.

Specifications

Product Line
Ion Torrent™
For Use With (Application)
Comparative Sequencing (Germline Mutations - 50:50)
For Use With (Equipment)
Ion 550 Chip, Ion 550 Kit-Chef
Product Type
Single Chip Supplemental Kit
Shipping Condition
Approved for shipment at Room Temperature or on Wet Ice

Contents & Storage

4 Ion S5 Chef Supplies, store at room temperature
1 Ion S5 Sequencing Solutions, store at room temperature
1 Ion S5 Sequencing Reagents, store at -20°C

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text