Applied Biosystems™

MagMAX CORE Nucleic Acid Purification Kit + Mechanical Lysis Module

Catalog number: A37487
Have Questions?
Applied Biosystems™

MagMAX CORE Nucleic Acid Purification Kit + Mechanical Lysis Module

Catalog number: A37487
Have Questions?
Catalog Number
A37487
Unit Size
100 tests
Price (USD)
Quantity
Catalog NumberUnit SizePrice (USD)Quantity
A37487100 tests
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
This MagMAX CORE Mechanical Lysis Module Bundle includes a MagMAX CORE Mechanical Lysis Module and MagMAX CORE Nucleic Acid Purification Kit. The Mechanical Lysis Module is also available separately.

The Applied Biosystems MagMAX CORE sample preparation solution is a universal sample preparation system designed to meet current and future veterinary testing needs. This modular technology is optimized for the purification of nucleic acids from a wide range of sample matrices commonly submitted for veterinary diagnostic testing. The MagMAX CORE Mechanical Lysis Module is offered as a supplement to the MagMAX CORE Nucleic Acid Purification Kit to aid in the processing of hard-to-lyse organisms like Mycobacterium avium paratuberculosis (MAP) and bovine tuberculosis in feces, milk, and environmental samples.

Features include:
Convenience—a complete solution using the same CORE chemistry
Confidence—higher-quality nucleic acids from difficult-to-lyse samples, including both small (0.3 g) and large (2–5 g) volume feces samples
Flexible—purchase only what you need to lyse MAP and other hard-to-lyse organisms

The MagMAX CORE Nucleic Acid Purification Kit and the MagMAX CORE Mechanical Lysis Module help make your lab more efficient through greater convenience, consistent performance, and time and cost savings.

Specifications

Sample Type
Bacteria, Blood, Buccal Samples, Cells, DNA, RNA, Tissue, Virus
For Use With (Application)
Next-Generation Sequencing, RT-PCR (Endpoint), Real-Time PCR
High-throughput Compatibility
Automated Protocols, Manual Protocols, Multiplexing
Product Line
MagMAX™
Quantity
100 Tests
Shipping Condition
Room Temperature
Target
Total RNA, Pathogen DNA⁄RNA, Genomic DNA, Viral DNA⁄RNA
Unit Size
100 tests

Contents & Storage

• 1 x 50 mL MagMAX CORE Lysis Solution
• 1 x 45 mL MagMAX CORE Binding Solution
• 1 x 60 mL MagMAX CORE Wash Solution 1
• 1 x 60 mL MagMAX CORE Wash Solution 2
• 1 x 12 mL MagMAX CORE Elution Buffer
• 1 x 2.2 mL MagMAX CORE Magnetic Beads
• 1 x 1.25 mL MagMAX CORE Proteinase K
• 1 x 45 mL MagMAX CORE Clarifying Solution
• 100 MagMAX CORE Bead Beating Tubes (2 mL)

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text