You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
Home
Shop All Products
Molecular Biology Reagents and Kits
DNA Extraction and Purification
MagMAX™ CORE Glass Microbeads
Applied Biosystems™
MagMAX™ CORE Glass Microbeads
Catalog number:
A37489
Related applications:
Viral RNA⁄DNA Purification
Have Questions?
Contact Us
Applied Biosystems™
MagMAX™ CORE Glass Microbeads
Catalog number:
A37489
Related applications:
Viral RNA⁄DNA Purification
Have Questions?
Contact Us
Catalog Number
A37489
Unit Size
100 tests
Price (USD)
47.84
Quantity
Save to list
Add to cart
Catalog Number
Unit Size
Price (USD)
Quantity
A37489
100 tests
47.84
Save to list
Add to cart
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
MagMAX CORE Glass Microbeads are offered as a supplement to the
MagMAX CORE Nucleic Acid Purification Kit
and the
MagMAX CORE Mechanical Lysis Module
. The beads aid in the homogenization of completely dessicated feces such as environmental samples or fecal pellets.
For a kit that contains these microbeads as well as the MagMAX CORE Nucleic Acid Purification Kit and the MagMAX CORE Mechanical Lysis Module, please see
Cat. No. A37488
.
Specifications
Sample Type
Blood, Buccal Samples, Cells, DNA, RNA, Tissue, Virus
For Use With (Application)
Next-Generation Sequencing, RT-PCR (Endpoint), Real-Time PCR
High-throughput Compatibility
Manual Protocols, Multiplexing
Product Line
MagMAX™
Shipping Condition
Room Temperature
Target
Total RNA, Pathogen DNA⁄RNA, Genomic DNA, Viral DNA⁄RNA
Contents & Storage
50 mL bottle, store at room temperature
Figures
Documents & Downloads
Certificates
Search by lot number or partial lot number
Search
Request a Certificate
Frequently asked questions (FAQs)
Citations & References
Search citations by name, author, journal title or abstract text
Search