MCF7 (+hIGF-1) Control Lysate (1 mg/mL)
MCF7 (+hIGF-1) Control Lysate (1 mg/mL)
Thermo Scientific™

MCF7 (+hIGF-1) Control Lysate (1 mg/mL)

Thermo Scientific™ MCF7 (+hIGF-1) Control Lysate is a stimulated cell lysate that has been validated for the expression levels and phosphorylation of key targets in the AKT signaling pathway.
Have Questions?
Catalog NumberQuantity
A379590.5 mL
Catalog number A37959
Price (USD)
392.70
Special offer
Online exclusive
Ends: 15-Aug-2025
462.00
Save 69.30 (15%)
Each
In stock
Add to cart
Quantity:
0.5 mL
Price (USD)
392.70
Special offer
Online exclusive
Ends: 15-Aug-2025
462.00
Save 69.30 (15%)
Each
Add to cart
Ask our AI about this Product
Thermo Scientific™ MCF7 (+hIGF-1) Control Lysate is a stimulated cell lysate that has been validated for the expression levels and phosphorylation of key targets in the AKT signaling pathway.
  • Validated for the detection and phosphorylation of AKT, PTEN, IRS1, IGF1-R, GSK3a, GSK3b, PRAS40, mTOR, p70S6K, and TSC2 proteins
  • Optimized - stimulation and lysis conditions optimized for detection of AKT signaling targets
  • Convenient - lysate is ready-to-use for immunoprecipitation, immunoprecipitation to mass spectrometry, and western blotting applications
  • Concentration: 1 mg/mL
  • Stable for two years at -80°C
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodMass Spectrometry
Final Product TypeProtein
For Use With (Equipment)Mass Spectrometer
Quantity0.5 mL
Shipping ConditionDry Ice
Starting MaterialCell Lysate
Workflow StepSample Enrichment
FormFrozen Liquid
Product LineThermo Scientific™
TypeControl Lysate
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Frequently asked questions (FAQs)

What are the characteristics of MCF7 (+hIGF-1) Control Lysate, and what is its main purpose?

MCF7 (+hIGF-1) Control Lysate is a stimulated MCF-7 cell lysate that has been validated for the expression levels and phosphorylation of key targets (AKT, PTEN, IRS1, IGF1-R, GSK3a, GSK3b, PRAS40, mTOR, p70S6K, and TSC2) in the AKT signaling pathway. It serves as a positive control for use with SureQuant AKT panel kits.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link