HCT 116 (+hIGF-1) Control Lysate (1mg/mL)
HCT 116 (+hIGF-1) Control Lysate (1mg/mL)
Thermo Scientific™
HCT 116 (+hIGF-1) Control Lysate (1mg/mL)
Thermo Scientific HCT 116 (+hIGF-1) Control Lysate is a stimulated cell lysate that has been validated for the expression levelsRead more
Have Questions?
Catalog NumberQuantity
A379610.5 mg (1 mg/mL)
Catalog number A37961
Price (USD)
383.35
Special offer
Online exclusive
Ends: 15-Aug-2025
451.00
Save 67.65 (15%)
Each
-
Add to cart
Quantity:
0.5 mg (1 mg/mL)
Price (USD)
383.35
Special offer
Online exclusive
Ends: 15-Aug-2025
451.00
Save 67.65 (15%)
Each
Add to cart
Ask our AI about this Product
Thermo Scientific HCT 116 (+hIGF-1) Control Lysate is a stimulated cell lysate that has been validated for the expression levels and phosphorylation of key targets in the RAS signaling pathway.
HCT 116 (+hIGF-1) Control Lysate features include:
• Validated for the expression RAS by western blotting and of NRAS, KRAS, and HRAS by mass spectrometry analysis
• Optimized—stimulation and lysis conditions optimized for detection of RAS signaling targets
• Convenient—lysate is ready-to-use for immunoprecipitation, immunoprecipitation to mass spectrometry, and western blotting applications
• Stable for two years at -80°C
The RAS family of GTPase proteins modulates two key signaling pathways involved in cell growth and proliferation: the MAPK and PI3K pathways. Aberrant activation of RAS proteins promotes constitutive activation of cell growth and proliferation that can result in cancer. Mutations in the NRAS, KRAS, and HRAS oncogenes are the most common mutations in human cancer. This lysate has been validated for expression levels of the three isoforms of RAS using mass spectrometry.
The HCT 116 cell line is a human colon cancer epithelial cell line and has been used for proliferation and tumorigenicity studies for colon cancer. The HCT 116 (+hIGF-1) cell lysate is provided as frozen lysate at a standard concentration of 1 mg/mL (500 mg per vial). Lysates are prepared in non-denaturing buffer without loading dyes or dissociating agents (i.e., no 2-mercaptoethanol or dithiothreitol has been added). For western blot analysis, heat lysate to 95°C for 5 minutes. Add reducing agent before heating if desired. The recommended loading volume per lane is 10–20 μL (10–20 μg).
Related products
SureQuant AKT Pathway (Phospho) Multiplex Panel (Absolute Quantitation)
SureQuant RAS Isoform Panel (Absolute Quantitation)
SureQuant TP53 Panel (Absolute Quantitation)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodMass Spectrometry
Final Product TypeProtein
For Use With (Equipment)Mass Spectrometer
Quantity0.5 mg (1 mg/mL)
Shipping ConditionDry Ice
Starting MaterialCell Lysate
Workflow StepSample Enrichment
FormFrozen Liquid
Product LineThermo Scientific™
TypeControl Lysate
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Frequently asked questions (FAQs)
What are the characteristics of HCT 116 (+hIGF-1) Control Lysate, and what is its main purpose?
HCT 116 (+hIGF-1) Control Lysate is a stimulated HCT 116 cell lysate that has been validated for the expression levels and phosphorylation of key targets (NRAS, KRAS, and HRAS) in the RAS signaling pathway. It serves as a positive control for use with SureQuant RAS panel kits.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link