BT-549 Control Lysate (1mg/mL)
BT-549 Control Lysate (1mg/mL)
Thermo Scientific™
BT-549 Control Lysate (1mg/mL)
Thermo Scientific BT-549 Control Lysate is a stimulated cell lysate that has been validated for the expression levels and phosphorylationRead more
Have Questions?
Catalog NumberQuantity
A379620.5 mg (1 mg/mL)
Catalog number A37962
Price (HKD)
3,743.00
Each
Add to cart
Quantity:
0.5 mg (1 mg/mL)
Price (HKD)
3,743.00
Each
Add to cart
Ask our AI about this Product
Thermo Scientific BT-549 Control Lysate is a stimulated cell lysate that has been validated for the expression levels and phosphorylation of key targets in the TP53 signaling pathway.
BT-549 Control Lysate features include:
• Validated for the expression of TP53 and phosphorylated TP53 by western blotting and mass spectrometry analysis
• Optimized—stimulation and lysis conditions optimized for detection of TP53 signaling targets
• Convenient—lysate is ready-to-use for immunoprecipitation, immunoprecipitation to mass spectrometry, and western blotting applications
• Stable for two years at -80°C
The tumor suppressor protein TP53 functions as a negative checkpoint regulator for abnormal cell signaling, and can induce cell cycle arrest, activate DNA repair proteins, or initiate apoptosis. Mutations in TP53 often result abnormal cell growth and are commonly found in many types of cancer. This lysate has been validated for TP53 expression and phosphorylation.
The BT-549 cell line is a human triple negative epithelial breast cancer cell line derived from mammary gland. The BT-549 cell lysate is provided as frozen lysate at a standard concentration of 1 mg/mL (500 mg per vial). Lysates are prepared in non-denaturing buffer without loading dyes or dissociating agents (i.e., no 2-mercaptoethanol or dithiothreitol has been added). For western blot analysis, heat lysate to 95°C for 5 minutes. Add reducing agent before heating if desired. The recommended loading volume per lane is 10–20 μL (10–20 μg).
Related products
SureQuant AKT Pathway (Phospho) Multiplex Panel (Absolute Quantitation)
SureQuant RAS Isoform Panel (Absolute Quantitation)
SureQuant TP53 Panel (Absolute Quantitation)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodMass Spectrometry
Final Product TypeProtein
For Use With (Equipment)Mass Spectrometer
Quantity0.5 mg (1 mg/mL)
Shipping ConditionDry Ice
Starting MaterialCell Lysate
Workflow StepSample Enrichment
FormLiquid
Product LineThermo Scientific™
TypeControl Lysate
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link