Ion Torrent™
Ion GeneStudio™ S5 System with SmartStart
Catalog number: A38405
Have Questions?
Ion Torrent™
Ion GeneStudio™ S5 System with SmartStart
Catalog number: A38405
Have Questions?
Catalog Number
A38405
Unit Size
System + SmartStart
Price (USD)
Catalog Number
Unit Size
1 system
Price (USD)
Catalog NumberUnit SizePrice (USD)
A38405System + SmartStartRequest A Quote
A381941 systemRequest A Quote
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Ion GeneStudio S5 System is a semiconductor-based next-generation sequencing (NGS) system that enables simple targeted sequencing workflows. The Ion GeneStudio S5 System is optimized to provide affordability for labs focused on smaller panels and lower sequencing throughput requirements. With cartridge-based reagents, the system is simple to use and offers scalability and flexibility. It uses Ion 5 Series chips to support a broad range of high-throughput sequencing for clinical research and research applications from microbial genomes and gene panels to exomes and transcriptomes.
Key features:
• Select the chip (Ion 510–540 chip) that fits your application and throughput needs to achieve from 2–80 million reads per run
• Reduce setup time and complexity with cartridge-based plug-and-play reagents
• Complete run and analysis in hours and move forward with your experiments
The Ion GeneStudio S5 Series
The Ion GeneStudio S5 Series sequencers share the same foundational technology and fluidic architecture across all three instruments: this Ion GeneStudio S5 System, the Ion GeneStudio S5 Plus System, and Ion GeneStudio S5 Prime System. The primary difference across the three instruments is on-sequencer analysis time.
Speed
The Ion GeneStudio S5 Series leverages the speed of semiconductor sequencing to enable the production of high quality sequencing data in a few hours and enables you to go from DNA library to data in as little as 24 hours with only 45 minutes of total hands-on time when paired with the Ion Chef System.
Simplicity
With single-day installation, a straightforward user interface, and cartridge-based reagents, the Ion GeneStudio S5 Series sequencers are easy to use. Combined with Ion AmpliSeq technology for target selection, the Ion Chef System for automated library and template preparation, and Ion Reporter Software for automated variant annotation, targeted sequencing is simpler than ever.
Small sample input
Low input DNA from challenging samples types such as formalin-fixed paraffin-embedded (FFPE) tissue, retrospective samples from fine needle aspirates, and cell-free DNA extracted from blood (cfDNA) can be difficult to sequence on competitor next-generation sequencers. However, with Ion AmpliSeq technology and the Ion GeneStudio S5 Series, you can use as little as 1 ng input DNA or RNA.
Scalability
Why buy multiple systems for your lab when you can run all of your targeted sequencing or microbial applications on a single next-generation sequencer? The Ion 5 Series chips that run on the Ion GeneStudio S5 Series provide several throughput options, so your lab has the flexibility to scale from small to large projects, enabling multiple targeted sequencing applications on a single system.
Simple analysis and storage solutions
Torrent Suite Software and Ion Reporter Software make NGS easy for beginners and experts alike. Plan, monitor, track, and analyze your runs using Torrent Suite Software (pre-installed on the Torrent Server). Integrate, annotate, and interpret variants using Ion Reporter Software (Thermo Fisher Cloud or local options available based on your needs).
Services and support
With more than 2,000 field personnel, Thermo Fisher Scientific has the most extensive network of dedicated Field Application Specialists and Service Engineers to work with you, so you can be confident that we will be there when and where you need us most.
Service and support plans—comprehensive plans to maximize instrument performance and ensure availability of critical systems
Qualification services—document and ensure instrument is installed, operating, and performing to manufacturer’s specifications
Training—courses for experimental design, data analysis, troubleshooting, applications, and instrument operation
Bioinformatics—tailored support for NGS informatics execution, software use, data analysis, and downstream applications
DataSafe Solution—storage and back-up strategy, design, hardware, and support for all lab data
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product Line
GeneStudio™
Software Type
Torrent Suite Software, Ion Reporter Software
Shipping Condition
Room Temperature
Package Components
Catalog Number:
A38194
Unit Size:
1 system
Qty:
1
Catalog Number:
TRN00230
Unit Size:
Ion FAS On-Site Training, 1 day
Qty:
1
ZGPCSCBFX10
Catalog Number:
ZGPCSCBFX10
Unit Size:
Qty:
1
A33125
Catalog Number:
A33125
Unit Size:
Qty:
1