Thermo Scientific™

Pro-Detect™ Rapid Myc Competitive Assay Kit

Catalog number: A38512
Have Questions?
Thermo Scientific™

Pro-Detect™ Rapid Myc Competitive Assay Kit

Catalog number: A38512
Have Questions?
Catalog Number
A38512
Unit Size
10 assays
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A3851210 assays
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Thermo Scientific Pro-Detect Rapid Myc Competitive Assay Kit is a single-use, dipstick lateral flow kit for quick, easy detection of Myc (sequence: EQKLISEEDL)-tagged proteins in protein expression and purification systems. This lateral flow assay uses membrane-based strips coated with dye-bound conjugated antibodies specific to Myc tag to provide a qualitative detection within ten minutes. This assay can be used to detect Myc-tagged proteins directly from cell culture media (supernatant) or cell lysates without any special instrumentation or handling.

Features of the Pro-Detect Rapid competitive assay kits:
Easy to adopt—lateral flow strips can be used early in and throughout expression to monitor and confirm expression of desired protein-tagged target
Simple, fast method—simply dilute sample using provided diluent and dip in the lateral flow strip for qualitative results within 5-10 minutes
Sensitive—detect protein tags in supernatant or cell lysate samples down to low μg/mL concentrations
Specific—assay does not cross-react with common lysis and protein preparation reagents
Reliable—results are similar to other antibody immunoassays, including western blot and ELISA
Long shelf life—stable for at least one year at 4°C

Each Pro-Detect Rapid competitive assay is performed by simply adding a properly diluted tissue culture supernatant, lysate, or purified protein sample to a test tube or microplate well and dipping in a lateral flow strip. Gold-conjugated detection antibodies embedded in the strip form complexes with the tagged protein in the sample. In the presence of target protein tag, the dye-bound conjugated antibodies will bind to the tagged protein present in the sample instead of the protein tag bound at the test lines. Therefore, in a positive result, a competitive lateral flow assay will show decreasing intensity of the visible test lines until all test lines have disappeared. When there is no target protein tag present, the dye-bound conjugated antibodies specific to the target will bind at the test line(s), resulting in visual test lines in a negative result.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detectable Analytes
Myc (EQKLISEEDL)-tagged proteins
Assay
Lateral Flow Rapid Assay
Detection Method
Chromogenic
Product Line
Pro-Detect™
Product Type
Assay
Specificity
Protein tag
Sufficient For
10 Assays
For Use With (Application)
Lateral Flow Rapid Assay
Quantity
10 Assays
Shipping Condition
Wet Ice
Target
Protein

Contents & Storage

• 10 Rapid Assay Strips
• Sample Diluent, 15 mL

Sufficient for 10 assay determinations

Store at 2–8°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text