AutoLys M Tubes are for the preparation of FFPE samples for nucleic acid extraction. They feature a novel tube-in-tube designRead more
Have Questions?
Catalog Number
Quantity
A38738
25 each
Catalog number A38738
Price (USD)
172.00
Each
In stock
Add to cart
Quantity:
25 each
Price (USD)
172.00
Each
Add to cart
AutoLys M Tubes are for the preparation of FFPE samples for nucleic acid extraction. They feature a novel tube-in-tube design that creates cleared lysates from FFPE tissue samples, without the need for deparaffinization or wash steps, while increasing DNA and RNA yields. Just add your FFPE sample to the tube, followed by protease buffer and enzyme from the MagMAX FFPE DNA/RNA Ultra Kit. Once digestion is complete, lift the tubes from the AutoLys M Tube Rack using the AutoLys M TubeLifter or AutoLys M Tube Pliers and spin down the samples using a benchtop centrifuge with plate holder (SBS standard format). The FFPE-associated wax and debris are held in the upper chamber of the tube while the lysate passes through. Nucleic acids can then be purified from the clarified lysate using the MagMAX FFPE DNA/RNA Ultra Kit.
Features of the AutoLys M Tube system include: • FFPE sample lysis and nucleic acid separation from debris in a single tube • 2-D barcode on bottom tube • Elimination of use of organic solvents (xylene, CitriSolv) for deparaffinization and washes • Reduced processing time • Increased nucleic acid yield
The AutoLys M Tube system is a unique way to process your FFPE samples that will result in improved nucleic acid yields when used in conjunction with the MagMAX FFPE DNA/RNA Ultra Kit, helping ensure the success of downstream assays and reducing the time to data analysis.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)AutoLys TubeLifter System
Quantity25 each
Shipping ConditionRoom Temperature
Sample TypeFFPE Tissue
Unit SizeEach
Contents & Storage
• 25 AutoLys M tubes • 25 AutoLys M caps
Frequently asked questions (FAQs)
Can I use a heat block for the AutoLys M tube incubation?
No. The AutoLys M tubes don't fit in a standard 1.5 mL or 2.0 mL heat block. We recommend incubating the tubes in the tube racks in an incubator or oven.
What kind of centrifuge do I need to spin down the AutoLys M Tubes (Cat. No. A38738)?
Any centrifuge with plate adapters will work. The AutoLys M Tube Racks fit nicely into those adapters.
Do I need to buy the AutoLys M TubeLifter (Cat. No. A37956) or AutoLys M Tube Pliers (Cat. No. A38261) to use the AutoLys M Tubes (Cat. No. A38738)?
No, the AutoLys M Tubes can be lifted manually, but the tools significantly reduce hand strain and increase ease of use as well as throughput.
Do the AutoLys M Tubes come with the MagMAX FFPE DNA/RNA Ultra Kit?
No, they have to be purchased separately together with some other products that are necessary for using the AutoLys M Tube system efficiently:
- AutoLys M Tubes and Caps (Cat. No. A38738)
- Autolys M Tube Locking Lid (Cat. No. A37954)
- AutoLys M TubeLifter (Cat. No. A37956)
- AutoLys M Tube Pliers (Cat. No. A38261)
- AutoLys M Tube Racks (Cat. No. A37955)
How does using the the AutoLys M Tube system help in increase the DNA/RNA yield from FFPE samples?
The FFPE-associated wax and debris are held in the upper chamber of the AutoLys M tube while the lysate passes through. Nucleic acids can then be purified from the clarified lysate. The AutoLys M tubes thus reduce manipulation of the sample eliminating the need to pellet the tissue and wash, so tissue loss is minimized