SeqScape™ Software v4.0, Initial License
Product Image
Applied Biosystems™
SeqScape™ Software v4.0, Initial License
This is an Initial License for SeqScape Software. SeqScape Software is a resequencing package designed for mutation detection and analysis,Read more
Have Questions?
Catalog number A38880
Price (USD)/ 1 license
-
This is an Initial License for SeqScape Software. SeqScape Software is a resequencing package designed for mutation detection and analysis, SNP discovery and validation, pathogen sub-typing, allele identification, and sequence confirmation. It provides library functions for comparison to a known group of sequences, as well as 21 CFR Part 11 functionalities.
Register to download a previous Windows 7-compatible trial version of this software ›
Specifications
Product LineSeqScape™
Software TypeSequencing Data Analysis
Data EntryCE Sequencing (.ab1 files)
LicenseInitial
Operating SystemWindows 10 Pro/IoT, Windows 7 Pro
Product TypeSeqScape Software V4.0 Initial License
Quantity1 license
Software CategorySupports files from all AB Genetic Analyzers
Unit Size1 license
Contents & Storage
Software/CD