Variant Reporter™ Software v3.0, Initial License
Product Image
Applied Biosystems™
Variant Reporter™ Software v3.0, Initial License
This is the Initial License for Variant Reporter Software. This software performs comparative sequencing, also known as direct sequencing, medicalRead more
Have Questions?
Catalog NumberQuantity
A388841 license
Catalog number A38884
Price (USD)
-
Quantity:
1 license
Ask our AI about this Product
This is the Initial License for Variant Reporter Software. This software performs comparative sequencing, also known as direct sequencing, medical sequencing, PCR sequencing, and resequencing with DNA sequencing files. The software is designed for reference-based and non-reference-based analysis such as mutation detection and analysis, SNP discovery and validation, and sequence confirmation. The robust algorithms will call SNPs, mutations, insertions, deletions, and heterozygous insertions/deletions for data generated using the Applied Biosystems genetic analyzers.
Specifications
Data EntryCE Sequencing (.ab1 files)
LicenseInitial
Operating SystemWindows 10 Pro/IoT, Windows 7 Pro Service Pk. 1
Product TypeVariant Reporter Software V3.0 Initial License
Quantity1 license
Software CategorySupports files from all AB Genetic Analyzers
Product LineVariant Reporter™
Software TypeSequencing Data Analysis
Unit SizeEach
Contents & Storage
Software/CD
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
0 results displayed, search above for a specific certificate
Frequently asked questions (FAQs)
Variant Reporter Software v3.0 fully supports data generated from the following instruments running all versions of Data Collection Software:
- Applied Biosystems SeqStudio Genetic Analyzer
- Applied Biosystems 3500 Genetic Analyzer
- Applied Biosystems 3500xL Genetic Analyzer
- Applied Biosystems 3130 Genetic Analyzer
- Applied Biosystems 3130xl Genetic Analyzer
- Applied Biosystems 3730 DNA Analyzer
- Applied Biosystems 3730xl DNA Analyzer
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.
The primary updates in Variant Reporter Software v3.0 are the following:
- Variant Reporter Software v3.0 supports installation on Microsoft Windows 7 (64-bit and 32-bit) and Microsoft Windows 10 Professional (64-bit) operating systems.
- Variant Reporter Software v3.0 uses KB Basecaller version 1.4.2.4 for basecalling.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.
For importing data, there are many ways to do so:
More information on backing up the Data Store can be found in the Variant Reporter Help Contents under the keyword “Back-up”.
1. Select the Data Store where you would like the data imported to. To choose a Data Store:
a. Click the Manage button in the Data Store section of the Dashboard View.
b. Highlight the Data Store you wish to use and click on Change Location to select it. Click Close to close the window.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
To archive a Variant Reporter software project from the Dashboard:
1. Open the Dashboard View.
2. In the Project tab select the project you want to archive, then click .
3. In the Archive Destination dialog box, browse to the folder where you want the project archived.
4. Click OK. The project file you archived will no longer appear in the Variant Reporter Dashboard.
More information on archiving Project data can be found in the Variant Reporter Help Contents under the keyword “Archive”.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
To back up the Data Store:
1. Open the Dashboard View.
2. Click .
3. In the Browse for Back-up Folder Destination dialog box, select the folder that you want to back up the Data Store to, then click OK. Your project will start to get backed up.
Note: All associated Projects, Analysis Parameters, and References contained in the current Data Store will be copied to the folder you selected when you back up the Variant Reporter Data Store.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
Share catalog number, name or link
1x1 image pixel for data collection