ExpiFectamine™ Sf Transfection Reagent
Detect rhabdovirus contamination in Sf9 cultures. Learn more »
ExpiFectamine™ Sf Transfection Reagent
Gibco™

ExpiFectamine™ Sf Transfection Reagent

ExpiFectamine Sf Transfection Reagent is a next-generation cationic lipid-based reagent for efficient DNA transfection of insect cells using fast, flexibleRead more
Have Questions?
Catalog number A38915
Price (USD)/ 1 mL
642.65
Online exclusive
654.00 
Save 11.35 (2%)
-
Add to cart
Price (USD)/ 1 mL
642.65
Online exclusive
654.00 
Save 11.35 (2%)
Add to cart
ExpiFectamine™ Sf Transfection Reagent
Catalog numberA38915
Price (USD)/ 1 mL
642.65
Online exclusive
654.00 
Save 11.35 (2%)
-
Add to cart
ExpiFectamine Sf Transfection Reagent is a next-generation cationic lipid-based reagent for efficient DNA transfection of insect cells using fast, flexible protocols.

ExpiFectamine Sf Transfection Reagent:
• Delivers robust and reproducible transfection, giving you greater confidence in your results
• Enables transfection of plasmid DNA in adherent and suspension Sf9 and Sf21 cell cultures for reliable recombinant baculovirus production
• Facilitates efficient production of high-titer, high-quality P0 recombinant baculovirus in suspension culture format without the need for further virus amplification
• Exhibits low toxicity and does not require medium change post-transfection
• Is part of a simple, fast protocol with a short 5-minute complexation time
• Demonstrates linear scalability of transfections from culture volumes of 4 mL to 500 mL in suspension format

The ExpiFectamine Sf Transfection Reagent is a core component of the ExpiSf Expression System and has been optimized for transfection of ExpiSf9 Cells. However, the reagent also demonstrates superior performance using other Sf9 and Sf21 insect cells cultured in serum-free media such as Sf-900 II SFM or Sf-900 III SFM in both adherent and suspension culture formats.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeInsect Cells
Sample TypeBaculovirus DNA, Plasmid DNA
Transfection TechniqueLipid-based Transfection
For Use With (Application)Transfection
High-throughput CompatibilityHigh-throughput Compatible
Product LineExpiFectamine™
Product TypeTransfection Reagent
Quantity1 mL
Serum CompatibleYes
Shipping ConditionApproved for shipment at Room Temperature or on Wet Ice
Unit Size1 mL
Contents & Storage
Store at 2–8°C, do not freeze

Shelf Life: 24 months from date of manufacture