Informational:Inquire about OEM or Commercial Supply version of this product here.
Applied Biosystems™

MagMAX™ Saliva gDNA Binding Beads

Catalog number: A39061
Have Questions?
Applied Biosystems™

MagMAX™ Saliva gDNA Binding Beads

Catalog number: A39061
Have Questions?
Catalog Number
A39061
Unit Size
45 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A3906145 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
These magnetic binding beads are a component of the MagMAX Saliva gDNA Isolation Kit. They are offered here as a stand-alone reagent for applications that require more magnetic binding beads than is provided in the kit.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Isolation Technology
Magnetic Bead
For Use With (Application)
Next-Generation Sequencing, PCR
High-throughput Compatibility
High-throughput Compatible, Manual Protocols, Automated Protocols
Product Line
MagMAX™
Product Type
Saliva gDNA Binding Beads
Quantity
45 mL
Shipping Condition
Room Temperature
Target
Genomic DNA

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text