Applied Biosystems™

MagMAX™ Saliva gDNA Lysis/Binding Solution

Catalog number: A39063
Have Questions?
Applied Biosystems™

MagMAX™ Saliva gDNA Lysis/Binding Solution

Catalog number: A39063
Have Questions?
Catalog Number
A39063
Unit Size
550 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A39063550 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
This Lysis/Binding solution is a component of the MagMAX Saliva gDNA Isolation Kit. It is offered here as a stand-alone reagent for applications that require more Lysis/Binding solution than is provided in the kit.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Application)
Next-Generation Sequencing, PCR
High-throughput Compatibility
High-throughput Compatible, Manual Protocols, Automated Protocols
Product Line
MagMAX™
Product Type
Saliva gDNA Lysis/Binding Solution
Quantity
550 mL
Shipping Condition
Room Temperature
Target
Genomic DNA

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text