Gibco™

pRABBIT IgG IRES-EmGFP Positive Control Vector

Catalog number: A39243
Have Questions?
Gibco™

pRABBIT IgG IRES-EmGFP Positive Control Vector

Catalog number: A39243
Have Questions?
Catalog Number
A39243
Unit Size
1 vial
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A392431 vial
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The pRABBIT IgG IRES-EmGFP Positive Control Vector is a mammalian expression control that expresses a complete, full-length rabbit IgG and EmGFP. It can be used as a positive expression control for transient expression systems such as the Expi293 Expression System, the ExpiCHO Expression System, the FreeStyle 293 Expression System, and the FreeStyle CHO Expression System.

Features of the pRABBIT IgG IRES-EmGFP Positive Control Vector include:
• A mix of pcDNA3.4 plasmids containing the coding sequences for EmGFP and the heavy and light chains of rabbit IgG
• Optimized mix of IgG light and heavy chain transfection-grade plasmids in a single, ready-to-use tube
• Expression of rabbit IgG at 450–500 mg/L in Expi293F cells
• Sufficient material for transfection of up to 150 mL of suspension culture in the Expi293, ExpiCHO, and FreeStyle systems

This control is included in the Expi293 GnTI-, Expi293 Inducible, and Expi293 Inducible GnTI- expression system kits.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Type
Positive Control DNA Vector
Shipping Condition
Dry Ice
Quantity
1 Vial
Grade
Transfection Grade

Contents & Storage

1 vial (1 mg/mL) pRABBIT IgG IRES-EmGFP Positive Control Vector, store at 2–8°C or long term storage at -20°C

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text