Tetracycline Hydrochloride
Tetracycline Hydrochloride
Gibco™

Tetracycline Hydrochloride

Tetracycline is a yellow, odorless, crystalline powder. It is commonly used as an agent to activate gene expression from anRead more
Have Questions?
Catalog NumberQuantity
A39246500 mg
Catalog number A39246
Price (EUR)
71,00
Each
-
Add to cart
Quantity:
500 mg
Price (EUR)
71,00
Each
Add to cart
Ask our AI about this Product
Tetracycline is a yellow, odorless, crystalline powder. It is commonly used as an agent to activate gene expression from an inducible expression plasmid in engineered cells expressing the tetracycline repressor protein. Tetracycline Hydrochloride is the hydrochloride salt of tetracycline.

CAS Number: 64-75-5
Formula: C22H24N2O2HCl (molecular weight: 480.91)
Solubility: Soluble in water, DMSO, methanol, and ethanol. For cell culture applications, we recommend reconstituting in water.

Tetracycline Hydrochloride is supplied in a glass vial containing 500 mg with 96% purity.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Recommended StorageStore at room temperature.

Shipping ConditionWet Ice
Physical FormPowder
Quantity500 mg
TypeTetracycline Hydrochloride
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link