Thermo Scientific™

EZ-Link™ Sulfo-NHS-Biotin, No-Weigh™ Format

Catalog number: A39256
Have Questions?
Thermo Scientific™

EZ-Link™ Sulfo-NHS-Biotin, No-Weigh™ Format

Catalog number: A39256
Have Questions?
Catalog Number
A39256
Unit Size
10 x 1 mg
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
50 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A3925610 x 1 mg
-
2121750 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific EZ-Link Sulfo-NHS-Biotin is a short-chain, water-soluble biotinylation reagent for labeling antibodies, proteins and other molecules that have primary amines.

Features of EZ-Link Sulfo-NHS-Biotin:

Protein labeling—biotinylate antibodies to facilitate immobilization, purification or detection using streptavidin resins or probes
Cell surface labeling—biotinylates only surface proteins of whole cells because the negatively charged reagent does not permeate cell membranes
Amine-reactive—reacts with primary amines (-NH2), such as lysine side-chains or the amino-termini of polypeptides
Soluble—charged sulfo-NHS group increases reagent water solubility compared to ordinary NHS-ester compounds
Irreversible—forms permanent amide bonds; spacer arm cannot be cleaved
Very short—spacer arm (total length added to target) is 13.5 angstroms; it consists of the native biotin valeric acid group only.

Sulfo-NHS-Biotin is the shortest of three very similar EZ-Link Reagents that are water-soluble, non-cleavable, and enable simple and efficient biotinylation of antibodies, proteins and any other primary amine-containing macromolecules in solution. Specific labeling of cell surface proteins is another common application for these uniquely water-soluble and membrane impermeable reagents. Differing only in their spacer arm lengths, the three Sulfo-NHS-ester reagents offer the possibility of optimizing labeling and detection experiments where steric hindrance of biotin binding is an important factor. Sulfo-NHS-Biotin is offered in several package sizes and as complete protein labeling kits.

We manufacture biotin reagents to ensure the highest possible overall product integrity, consistency and performance for the intended research applications.

N-Hydroxysulfosuccinimide (NHS) esters of biotin are the most popular type of biotinylation reagent. NHS-activated biotins react efficiently with primary amino groups (-NH2) in alkaline buffers to form stable amide bonds. Proteins (e.g., antibodies) typically have several primary amines that are available as targets for labeling, including the side chain of lysine (K) residues and the N-terminus of each polypeptide.

Varieties of biotin NHS-ester reagents differ in length, solubility, cell permeability and cleavability. Non-sulfonated NHS-biotins are cell permeable but must be dissolved in organic solvent such as DMSO or DMF. Sulfo-NHS biotins (and those with pegylated spacers) are directly water soluble but not membrane permeable. Varieties containing disulfide bonds can be cleaved using reducing agents, enabling the biotin group to be disconnected from the labeled protein.

Related Products
EZ-Link™ Sulfo-NHS-Biotinylation Kit
EZ-Link™ Micro Sulfo-NHS-Biotinylation Kit
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Kit Contents
10 x 1 mg
Product Type
Sulfo-NHS-Biotin
Quantity
10 x 1 mg
Labeling Method
Chemical Labeling
Shipping Condition
Qualified for Ambient or Wet Ice
Product Line
EZ-Link™
Cell Permeability
Cell-Impermeant
Chemical Reactivity
Amine
Reactive Moiety
Sulfo-NHS Ester
Solubility
DMF (Dimethylformamide), DMSO (Dimethylsulfoxide), Water
Label or Dye
Biotin
Spacer
Short

Contents & Storage

Store desiccated at -20°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text