Thermo Scientific™

EZ-Link™ Sulfo-NHS-SS-Biotin, No Weigh Format

Catalog number: A39258
Have Questions?
Thermo Scientific™

EZ-Link™ Sulfo-NHS-SS-Biotin, No Weigh Format

Catalog number: A39258
Have Questions?
Catalog Number
A39258
Unit Size
10 x 1 mg
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
100 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A3925810 x 1 mg
-
21331100 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific EZ-Link Sulfo-NHS-SS-Biotin is a water-soluble, NHS-ester biotinylation reagent whose spacer arm includes a cleavable disulfide bond for reversible labeling of proteins and cell surface primary amines.

Features of EZ-Link Sulfo-NHS-SS-Biotin:

Protein labeling – biotinylate antibodies to facilitate immobilization, purification or detection
Cell surface labeling – biotinylates only surface proteins of whole cells because the negatively charged reagent does not permeate cell membranes
Amine-reactive—reacts with primary amines (-NH2), such as lysine side-chains, or the amino-termini of polypeptides
Cleavable—disulfide bond in spacer arm allows the biotin label to be removed using reducing agents such as DTT; only a small sulfhydryl group remains attached to the molecule
Soluble – charged sulfo-NHS group increases reagent water solubility compared to ordinary NHS-ester compounds
Medium length—spacer arm (total length added to target) is 24.3 angstroms; it consists of the native biotin valeric acid group extended by a 7-atom chain

Sulfo-NHS-SS-Biotin is a thiol-cleavable amine-reactive biotinylation reagent that contains an extended spacer arm to reduce steric hindrances associated with avidin binding. This reagent is water-soluble, enabling biotinylation to be performed in the absence of organic solvents such as DMSO or DMF for applications that cannot tolerate solvents or are complicated by their inclusion. The compound is particularly useful for labeling and purifying cell surface proteins, because (a) its sulfonate group prevents it from permeating cell membranes and (b) its cleavable spacer arm enables initially biotinylated proteins to be released from streptavidin affinity columns.

We manufacture biotin reagents to ensure the highest possible overall product integrity, consistency, and performance for the intended research applications.

N-Hydroxysulfosuccinimide (NHS) esters of biotin are the most popular type of biotinylation reagent. NHS-activated biotins react efficiently with primary amino groups (-NH2) in alkaline buffers to form stable amide bonds. Proteins (e.g., antibodies) typically have several primary amines that are available as targets for labeling, including the side chain of lysine (K) residues and the N-terminus of each polypeptide.

Varieties of biotin NHS-ester reagents differ in length, solubility, cell permeability and cleavability. Non-sulfonated NHS-biotins are cell permeable but must be dissolved in organic solvent such as DMSO or DMF. Sulfo-NHS biotins (and those with pegylated spacers) are directly water soluble but not membrane permeable. Varieties containing disulfide bonds can be cleaved using reducing agents, enabling the biotin group to be disconnected from the labeled protein.

Related Products
EZ-Link™ Sulfo NHS-SS Biotinylation Kit
EZ-Link™ Micro Sulfo-NHS-SS-Biotinylation Kit
Pierce™ Premium Grade Sulfo-NHS-SS-Biotin
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Type
Sulfo-NHS-SS-Biotin
Quantity
10 x 1 mg
Labeling Method
Chemical Labeling
Shipping Condition
Wet Ice
Product Line
EZ-Link™
Cell Permeability
Cell-Impermeant
Chemical Reactivity
Amine
Reactive Moiety
Sulfo-NHS Ester
Solubility
DMF (Dimethylformamide), DMSO (Dimethylsulfoxide), Water
Label or Dye
Biotin
Spacer
Cleavable, 24.3 Å

Contents & Storage

Store at -20°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text