The TrueMark OpenArray Urinary Tract Microbiota Comprehensive Plate is an efficient, easy-to-use OpenArray plate (56-assay format) for the characterization of key urinary tract microbial targets through real-time PCR. This plate contains TaqMan Gene Expression Assays for 17 microbial targets, plus Universal DNA Spike-In Positive Control and human RNase P RPPH1 control. The format of this OpenArray plate allows for three replicates to be run in parallel per plate for all microbial targets and two replicates for RNase P.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection Method
Primer-probe
GC-Rich PCR Performance
Low
Product Type
Urinary Tract Microbiota Comprehensive Plate
Quantity
1 plate
Shipping Condition
Approved for shipment at Room Temperature or on Wet Ice