Ponceau S Staining Solution
Ponceau S Staining Solution
Ponceau S Staining Solution
Ponceau S Staining Solution
Ponceau S Staining Solution
Thermo Scientific™

Ponceau S Staining Solution

Ponceaus S Staining Solution is a ready-to-use membrane stain for evaluating the transfer efficiency of a western blot.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A400002781000 mL
A40000279500 mL
A40000278X33 x 1000 mL
Catalog number A40000278
Price (USD)
111.65
Online Exclusive
117.00
Save 5.35 (5%)
1000 mL
Add to cart
Quantity:
1000 mL
Price (USD)
111.65
Online Exclusive
117.00
Save 5.35 (5%)
1000 mL
Add to cart
Ponceaus S Staining Solution is a ready-to-use membrane stain for evaluating the transfer efficiency of a western blot. This stain is compatible with nitrocellulose and PVDF membranes. Ponceau S staining is reversible and can be removed with a short incubation in 0.1% NaOH.
Ponceaus S Staining Solution is a ready-to-use membrane stain for evaluating the transfer efficiency of a western blot. This stain is compatible with nitrocellulose and PVDF membranes. Ponceau S staining is reversible and can be removed with a short incubation in 0.1% NaOH.

Features include:
• A nondestructive, reversible method to stain and detect proteins on nitrocellulose and PVDF membranes
• Minimal nonspecific interactions with the membrane surface; provides a reliable method for visualization of protein transfer to a membrane
• Simple and quick staining protocol that results in reddish-pink bands that are easily photographed
• Reversed easily in less than five minutes
• Subsequent western blot detection unaffected as the stain does not alter the membrane-bound protein

Specifications
Detection MethodColorimetric
For Use With (Application)Western Blotting
FormLiquid
Formulation0.1% (w/v) in 5% acetic acid
Product TypePonceau S Staining Solution
Quantity1000 mL
Unit Size1000 mL

Share catalog number, name or link