Search Thermo Fisher Scientific
Catalog Number | Quantity |
---|---|
A4000164002 | 100 L |
A4000164001 | 10 L |
Features of OptiPRO SFM AGT Medium include:
Serum-free, animal-origin-free, low-protein formulation
OptiPRO SFM is formulated with a very low protein concentration (< 10 μg/mL), for easier purification of recombinant proteins.
Easy adaptation for many popular cell lines
OptiPRO SFM was designed for growth of kidney-derived cell lines such as MDCK, MDBK, VERO, and PK-15. But it has also been used successfully to grow other cell lines, such as Hela, BHK-21, and COS-7, allowing the use of a single medium for multiple cell lines. OptiPRO SFM is formulated without L-glutamine for greater stability and extended shelf life. Simply add L-glutamine or Glutamax supplement as needed.
cGMP manufacturing and quality system
OptiPRO SFM is manufactured in a cGMP-compliant facility. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.
Scalability with Advanced Granulation Technology
Advanced Granulation Technology (AGT) helps simplify medium preparation at large scale with fast rehydration, a just-add-water protocol, and low dust generation, creating less pressure build-up during sterile filtration.
Share catalog number, name or link