Search
Search

| Catalog Number | Diameter (Metric) Inner | Length (Metric) |
|---|---|---|
| A40009234 | 20 cm | 20 cm |
| A40009249 | 10 cm | 20 cm |
| A40009226 | 14 cm | 20 cm |
POROS GoPure Benzyl chromatography columns are ready-to-use and designed specifically for use in clinical and commercial manufacturing. POROS GoPure Benzyl columns are prepared and packed in an ISO Class 7 environment designed to allow all product contact materials to meet USP <88> Class VI standards.
Each column is accompanied by column qualification documentation including bioburden and endotoxin. Packed with POROS™ Benzyl hydrophobic interaction resin, POROS GoPure Benzyl columns offer high binding capacity over a wide range of flow rates.
Features of POROS Benzyl resin include hydrophobic interaction chromatography for biomolecule purification. Through a benzyl functional group and enhanced biomolecule access via large pores, POROS Benzyl resin offers high dynamic capacity and high productivity.