Applied Biosystems™
Thermal Cycler Fleet Control Software
Catalog number: A40070
Have Questions?
Applied Biosystems™
Thermal Cycler Fleet Control Software
Catalog number: A40070
Have Questions?
Catalog Number
A40070
Unit Size
1 each
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A400701 each
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Applied Biosystems Thermal Cycler Fleet Control Software provides a new level of control and organization for the thermal cyclers in your lab. Connect to and manage any of your Applied Biosystems thermal cyclers through a central computer within your organization's firewall.
Features of Thermal Cycler Fleet Control Software include:
• Supplied pre-loaded on a Dell™ Latitude 3310 2-in-1 desktop computer. Mouse, keyboard, and standard 5-year warranty included.
• Access the software from any networked computer to quickly view, start, stop, and manage your thermal cycler fleet
• Manage run-reports on the network and automatically export them to a custom network location
• Create custom permissions to ensure that users, methods, and instruments are used according to your lab’s quality systems
• Connects to these compatible thermal cyclers: ProFlex PCR System, SimpliAmp Thermal Cycler, Veriti Thermal Cycler, MiniAmp thermal cyclers, and Automated Thermal Cycler (ATC)
Applied Biosystems PCR plastics
Applied Biosystems PCR plastics have been designed and validated to work with our thermal cyclers for more than 25 years. Options for every format and throughput need are available to ensure consistent, precise results for any experiment. To find the best match please visit our Plastics Selection Guide.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Voltage
100/240 V
Product Line
Applied Biosystems™
Display Type
19.5 in. Wide-Viewing Angle
Software Type
Instrument Mangement and Control
Product Type
Thermal Cycler Fleet Control Software
For Use With (Application)
PCR