Thermo Scientific™

SureQuant™ AKT Pathway (Phospho) Relative Quantitation Module

Catalog number: A40087
Have Questions?
Thermo Scientific™

SureQuant™ AKT Pathway (Phospho) Relative Quantitation Module

Catalog number: A40087
Have Questions?
Catalog Number
A40087
Unit Size
10 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A4008710 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Thermo Scientific SureQuant AKT Pathway (Phospho) Relative Quantitation Module provides a complete quantitation solution for the analysis of key phosphorylation events of AKT signaling pathway proteins using mass spectrometry (MS).

SureQuant AKT Pathway (Phospho) Relative Quantitation Module features include:
Complete—contains validated AQUA HeavyPeptide mix, diluent, matrix, system suitability standard including optimized acquisition and analysis methods
Validated—peptides rigorously tested for successful quantitation of each target peptide
Multiplex—able to quantitate phosphorylated AKT, PTEN, IRS1, IGF1-R, GSK3a, GSK3b, PRAS40, mTOR, p70S6K, and TSC2 in a single reaction

The AKT signaling pathway serves as a master regulatory pathway for cell growth and proliferation, and phosphorylation plays a major role in its modulation. Aberrant activation of this pathway promotes survival of tumor cells in many types of cancer. This multiplex kit is ideal for monitoring protein expression of ten key phosphorylated proteins in the AKT signaling pathway (AKT, PTEN, IRS1, IGF1-R, GSK3a, GSK3b, PRAS40, mTOR, p70S6K, and TSC2) in a single assay. The ability to simultaneously monitor and quantitate these targets in a single sample is a powerful tool for assessment of normal and abnormal pathway signaling.

The SureQuant AKT Pathway (Phospho) Relative Quantitation Module provides reagents for relative quantitation of 30 unique peptides from 10 phosphorylated AKT-mTOR signaling pathway target proteins using liquid chromatography (LC) and MS. AQUA Ultimate HeavyPeptide mixture is provided for use as a spike-in internal standard for relative quantitation in the multiplexed sample. Standardized methods and data process templates are provided for streamlined data acquisition and analysis.

The Pierce LC-MS/MS System Suitability Standard (7 x 5 Mix) is included in this module to assess dynamic range of LC-MS/MS systems before running the calibration curves and samples.

This kit can be used with the SureQuant AKT Pathway (Phospho) IP and MS Sample Preparation Module for enriched multiplex sample preparation. When used together, these modules provide a complete solution for sample preparation and analysis of the phosphorylated target proteins in the pathway.

Related products:
EASY-Spray LC Column
SureQuant AKT Pathway (Phospho) Multiplex Panel (Absolute Quantitation)
Pierce Trypsin Protease, MS Grade
Pierce LC-MS/MS System Suitability Standard

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method
Mass Spectrometry
For Use With (Equipment)
Thermo Scientific™ Q Exactive Mass Spectrometer, Mass Spectrometer
Shipping Condition
Approved for shipment on Wet or Dry Ice
Workflow Step
Target Selection
Product Line
SureQuant™
Quantity
1 Kit
For Use With (Application)
Mass Spectrometry
Type
Quantitation Module
Final Product Type
Peptides
Starting Material
Protein Samples
Sufficient For
10 Reactions

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text