Ion Torrent™

GX5™ Chip and Genexus™ Coupler

Catalog number: A40269
Have Questions?
Ion Torrent™

GX5™ Chip and Genexus™ Coupler

Catalog number: A40269
Have Questions?
Catalog Number
A40269
Unit Size
2 runs
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A402692 runs
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Ion Torrent GX5 Chip and Genexus Coupler includes two GX5 chips and two Genexus couplers, sufficient for two initializations of the Ion Torrent Genexus Integrated Sequencer. The GX5 Chip generates 12–15 million sequencing reads per lane, or 48–60 million reads per chip. The four-lane chip design provides maximum flexibility for sample processing, enabling sequencing of one to four lanes per run, with the in-use stability to utilize any remaining lanes within two weeks of the initial run.

As part of the Genexus Integrated Sequencer workflow, the GX5 Chip:
• Achieves 12–15 million reads per lane for 200–400 base-read libraries
• Maximizes flexibility with the ability to utilize one to four chip lanes per run
• Enables parallel processing of up to four compatible assays in a single run
• Supports intake variability of clinical research samples

The Ion Torrent Genexus Integrated Sequencer automates library preparation, sequencing, analysis, and reporting in a hands-off, walkaway workflow to deliver results in a single day. The innovative, multilane sequencing chip is designed to enable cost-effective processing of 1 to 32 single pool libraries.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment)
Genexus Integrated Sequencer
For Use With (Application)
Comparative Sequencing (Somatic Mutations - 10:50)
Green Features
Sustainable Disposal
Product Line
GX5™, Genexus™
Product Type
Coupler
Quantity
2 Runs
Shipping Condition
Room Temperature

Contents & Storage

• 2 GX5 chips
• 2 Genexus couplers

Store at room temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text