Ion Torrent™

Genexus™ GX5™ Starter Pack-AS

Catalog number: A40279
Have Questions?
Ion Torrent™

Genexus™ GX5™ Starter Pack-AS

Catalog number: A40279
Have Questions?
Catalog Number
A40279
Unit Size
32 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A4027932 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Ion Torrent Genexus GX5 Starter Pack for Ion AmpliSeq panels provides one set of all core consumables required to run the Ion Torrent Genexus Integrated Sequencer. This starter pack supports flexible, fully automated preparation and normalization of up to 32 Ion AmpliSeq libraries, as well as sequencing for up to eight chip lanes (two full chips). The Ion Torrent GX5 Chip is used in conjunction with this starter pack and must be ordered separately.

As part of the Genexus Integrated Sequencer GX5 workflow, the Genexus GX5 Starter Pack-AS:
• Automates library preparation, templating, and sequencing for 200–400 base-read Ion AmpliSeq libraries
• Supports intake variability of clinical research samples in a cost-effective manner
• Maximizes flexibility with the ability to sequence one to four chip lanes per run
• Enables parallel processing of up to four compatible assays in a single run
• Delivers fast and simple instrument set-up, with less than five minutes of hands-on time

The Ion Torrent Genexus Integrated Sequencer automates library preparation, sequencing, analysis, and reporting in a hands-off, walkaway workflow to deliver results in a single day. The innovative, multilane sequencing chip is designed to enable cost-effective processing of 1 to 32 single pool libraries.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment)
Genexus Integrated Sequencer
For Use With (Application)
Comparative Sequencing (Somatic Mutations - 10:50)
No. of Reactions
32
Product Line
GX5™, Genexus™
Product Type
Starter Pack-AS
Shipping Condition
Approved for shipment at Room Temperature or on Wet or Dry Ice

Contents & Storage

1 each of:
• Genexus Strip 2-AS, store at -10 to -20°C
• Genexus Strip 1, store at 2–8°C
• Genexus Barcodes 1-32 AS, store at room temperature
• Genexus Strip 4, store at -10 to -20°C
• Genexus Strip 3-GX5, store at 2–8°C
• Genexus Pipette Tips, store at room temperature
• Genexus Cartridge, store at -10 to -20°C
• Genexus Bottle 2, store at room temperature
• Genexus Bottles 1 and 3, store at room temperature

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text