Informational:Discover the future of Gibco Media selection with our upcoming updated experience. View details >
Gibco™

CTS™ LV-MAX™ Production Medium

Catalog number: A4124001
Have Questions?
Gibco™

CTS™ LV-MAX™ Production Medium

Catalog number: A4124001
Have Questions?
Catalog Number
A4124001
Unit Size
1 L
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
10 L
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
20 L
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
6 x 1 L
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A41240011 L
-
A412400210 L
-
A412400320 L
-
A41240046 x 1 L
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

Gibco CTS LV-MAX Production Medium is a chemically-defined, serum-free, protein-free medium validated for recombinant lentiviral vector production. In addition to supporting high cell density suspension cultures of CTS Viral Production Cells (HEK293F-derived cells), this medium is suitable for preclinical-, clinical-, and commercial-scale transient transfection and lentiviral production when used with the CTS LV-MAX Transfection Kit. CTS LV-MAX Production Medium undergoes stringent QC testing to ensure it is appropriate for the manufacturing of cell and gene therapy applications.

CTS LV-MAX Production Medium may be applicable for other recombinant viral vector production that uses a HEK293 medium.

CTS LV-MAX Production Medium features include:

  • Supports CTS Viral Production Cells to achieve densities of over 1 x 107 cells/mL with minimal cell clumping
  • Enables high viral titers of up to 1 x 108 TU/mL (unconcentrated lentiviral vectors–GFP)
  • Scalable protocol for 96-deep well blocks, flasks, and bioreactors
  • Regulatory-compliant medium manufactured in conformity with cGMP for medical devices, 21 CFR, Part 820, and certified to the ISO 13485. Drug Master File and Regulatory Support File available upon request.

CTS LV-MAX Production Medium is formulated with GlutaMAX-I reagent. It is ready to use, with no supplementation required. The chemically defined formulation results in high reproducibility and lot-to-lot reliability. The medium contains no human or animal-origin components. It is not recommended for adherent cell cultures. The formulation of this product is identical to research-grade LV-MAX Production Medium, but CTS LV-MAX Production Medium is validated for clinical and commercial therapeutic applications.

Custom configuration and packaging available upon request.

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.

Specifications

Cell Line
Viral Production Cells
Cell Type
HEK293
Classification
Animal Origin-free
Form
Liquid
Product Type
LV Production Medium
Shelf Life
24 months from date of manufacture
Sterility
Sterile-filtered
Format
Bottle
For Use With (Application)
LV Production
Manufacturing Quality
ISO 13485, MDSAP, FDA-registered, 21 CFR 820
Quantity
1 L
Shipping Condition
Ambient

Contents & Storage

1 L bottle. Store at 2°C to 8°C and protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text