Applied Biosystems™ RapidHIT™ ID System
Applied Biosystems™ RapidHIT™ ID System
Applied Biosystems™
Applied Biosystems™ RapidHIT™ ID System
The Applied Biosystems RapidHIT ID System is a fast and simple-to-use instrument that produces trusted lab-quality forensic DNA profiles inWeitere Informationen
Have Questions?
KatalognummerMenge
A418101 system
Katalognummer A41810
Preis (EUR)
-
Angebot anfordern
Menge:
1 system
Ask our AI about this Product
The Applied Biosystems RapidHIT ID System is a fast and simple-to-use instrument that produces trusted lab-quality forensic DNA profiles in as little as 90 minutes. The system integrates sample preparation and capillary electrophoresis to generate DNA profiles that are aggregated within Applied Biosystems RapidLINK Software for direct upload to the database of choice or for further review and analysis.
The RapidHIT ID System delivers the reliable, reproducible, high-quality data you have come to expect from the Applied Biosystems brand. This system is for use with Applied Biosystems RapidLINK Software.
Features of the Applied Biosystems RapidHIT ID System include:
• Sample to answer in as little as 90 minutes
• 1-minute hands-on time with integrated ACE and RapidIntel PLUS sample cartridges
• Consumables tracking through radio frequency identification (RFID)
• Intuitive touchscreen interface
• Facial recognition and barcode camera
• Fingerprint reader
• Up to twelve months of shelf life for both sample and primary cartridges
• FBI NDIS-approved rapid DNA system and booking station device*
* NDIS-approved for use with RapidHIT ID ACE GlobalFiler Express Sample Cartridge in accredited forensic DNA laboratories
Specifications
Menge1 system
VersandbedingungRaumtemperatur
ProduktlinieRapidHIT™
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Dokumente und Downloads
Zertifikate
Sicherheitsdatenblätter
Geben Sie die Katalognummer, den Namen oder Link frei.
1x1 image pixel for data collection