1x1 image pixel for data collection
Applied Biosystems™ RapidLINK™ Software v1.0 Familial Application
Applied Biosystems™ RapidLINK™ Software v1.0 Familial Application
Applied Biosystems™
Applied Biosystems™ RapidLINK™ Software v1.0 Familial Application
The Applied Biosystems RapidLINK Familial Application is an easy-to-use application software that generates a list of DNA profiles in theRead more
Have Questions?
Catalog NumberQuantity
A418191 annual license
Catalog number A41819
Price (USD)
-
Request A Quote
Quantity:
1 annual license
The Applied Biosystems RapidLINK Familial Application is an easy-to-use application software that generates a list of DNA profiles in the RapidLINK database that may be familiarly related to each other. The application generates a printable, summary report of profiles that may have a parent-child or sibling-sibling relationship to the profile being queried, and details the likelihood ratio of the overall relationship and the likelihood ratio of the allelic relationship at each locus. For use only with Applied Biosystems RapidLink Software v1.0.
Features of the RapidLINK Familial Application include:
• Identification of familiarly related DNA profiles
• Flexible with regard to size of pool
• Reduced workload for lab personnel
• Calculates likelihood ratios of parent-child and sibling-sibling relationships
• User-defined search stringency
• Automatically generated match report
Specifications
For Use With (Equipment)Applied Biosystems RapidLINK Software V1.0
Product TypeFamilial Application Software
Quantity1 annual license
Shipping ConditionRoom Temperature
Product LineRapidLINK™
Software TypeLicense
Unit SizeEach
Contents & Storage
1 document letter
Share catalog number, name or link