Informational:Discover the future of Gibco Media selection with our upcoming updated experience. View details >
Gibco™

BenchStable™ DMEM

Catalog number: A4192102
Have Questions?
Gibco™

BenchStable™ DMEM

Catalog number: A4192102
Have Questions?
Catalog Number
A4192102
Unit Size
10 x 500 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
500 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A419210210 x 500 mL
-
A4192101500 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

Gibco DMEM (Dulbecco's Modified Eagle Medium) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM include primary fibroblasts, neurons, glial cells, HUVECs, and smooth muscle cells, as well as cell lines such as HeLa, 293, Cos-7, and PC-12. We offer a variety of DMEM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

BenchStable DMEM has been engineered for storage at room temperature while supporting cellular proliferation and maximum cell densities comparable to conventional DMEM when supplemented with ≥10% fetal bovine serum (FBS).

This DMEM is modified as follows:
• Optimized for room temperature stability
• With High Glucose, GlutaMAX, Phenol Red
• Without Sodium Pyruvate

Using BenchStable DMEM
DMEM is unique from other media as it contains four times the concentration of amino acids and vitamins as the original Eagle's Minimal Essential Medium. DMEM was originally formulated with low glucose (1 g/L) and sodium pyruvate, but is often used with higher glucose levels, with or without sodium pyruvate. DMEM uses a sodium bicarbonate buffer system (3.7 g/L) and so requires a 5–10% CO2 environment to maintain physiological pH. DMEM with GlutaMAX supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format. BenchStable DMEM contains no proteins, lipids, or growth factors and so requires supplementation with ≥10% fetal bovine serum (FBS).

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Line
HeLa, 293, Cos-7, and PC-12
Classification
Animal Origin-free
Concentration
1 X
Form
Liquid
Product Type
DMEM (Dulbecco's Modified Eagle Medium)
Serum Level
Standard Serum Supplementation
Shelf Life
12 Months From Date of Manufacture
Sterility
Sterile-filtered
With Additives
High Glucose, GlutaMAX, Phenol Red
Without Additives
No Sodium Pyruvate, No HEPES
Green Features
Energy efficient, Fewer resources used, Sustainable packaging
Manufacturing Quality
cGMP-compliant under the ISO 9001: 2015 standard
Product Line
BenchStable™
Quantity
10 x 500 mL
Shipping Condition
Room Temperature
Unit Size
10 x 500 mL

Contents & Storage

Store at room temperature. Protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text