Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Compatible Ladders | E-Gel™ 1 Kb Plus Express Ladder |
Gel Percentage | 1% |
Gel Type | E-Gel |
Product Type | Agarose Gel |
Separation Range | 100 to 5000 bp |
Stain Type | SYBR™ Gold |
Wells | 22-well |
Quantity | 10 Gels |
Well Design | Double Comb |
Unit Size | Each |
Compatible Ladders | E-Gel™ 1 Kb Plus Express Ladder |
Gel Percentage | 1% |
Gel Type | E-Gel |
Product Type | Agarose Gel |
Separation Range | 100 to 5000 bp |
Stain Type | SYBR™ Gold |
Wells | 22-well |
Quantity | 2 x 10 Gels |
Well Design | Double Comb |
Unit Size | Each |
Compatible Ladders | E-Gel™ 1 Kb Plus Express Ladder |
Gel Percentage | 1% |
Gel Type | E-Gel |
Product Type | Agarose Gel |
Separation Range | 100 to 5000 bp |
Stain Type | SYBR™ Gold |
Wells | 22-well |
Quantity | 5 x 10 Gels |
Well Design | Double Comb |
Unit Size | Each |
Compatible Ladders | E-Gel™ 50 bp DNA Ladder |
Gel Percentage | 2% |
Gel Type | E-Gel |
Product Type | Agarose Gel |
Separation Range | 50 to 2000 bp |
Stain Type | SYBR™ Gold |
Wells | 22-well |
Quantity | 10 Gels |
Well Design | Double Comb |
Unit Size | Each |
Compatible Ladders | E-Gel™ 50 bp DNA Ladder |
Gel Percentage | 2% |
Gel Type | E-Gel |
Product Type | Agarose Gel |
Separation Range | 50 to 2000 bp |
Stain Type | SYBR™ Gold |
Wells | 22-well |
Quantity | 2 x 10 Gels |
Well Design | Double Comb |
Unit Size | Each |
Compatible Ladders | E-Gel™ 50 bp DNA Ladder |
Gel Percentage | 2% |
Gel Type | E-Gel |
Product Type | Agarose Gel |
Separation Range | 50 to 2000 bp |
Stain Type | SYBR™ Gold |
Wells | 22-well |
Quantity | 5 x 10 Gels |
Well Design | Double Comb |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Gel Percentage | Separation Range | Quantity | Price (USD) | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
A42345 | Each | 1% | 100 to 5000 bp | 10 Gels | 278.00 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A44887 | Each | 1% | 100 to 5000 bp | 2 x 10 Gels | 494.00 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A44888 | Each | 1% | 100 to 5000 bp | 5 x 10 Gels | 1,126.00 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A42346 | Each | 2% | 50 to 2000 bp | 10 Gels | 277.00 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A44889 | Each | 2% | 50 to 2000 bp | 2 x 10 Gels | 488.00 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A44890 | Each | 2% | 50 to 2000 bp | 5 x 10 Gels | 1,124.00 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Features of E-Gel EX Double Comb Agarose Gels include:
Ultra-sensitive detection of DNA
E-Gel EX Double Comb gels include SYBR Gold DNA stain and were developed for ultimate sensitivity, demonstrating up to five-fold greater sensitivity than comparable gels containing ethidium bromide. This superior sensitivity allows you to use lower amounts of sample, saving time and money.
Fast and convenient analysis
E-Gel EX Double Comb gels are our fastest-resolving agarose gels, enabling sample analysis in as little as 5-8 minutes. Each E-Gel EX Double Comb gel contains agarose, electrodes, SYBR Gold DNA stain, and a bufferless ion exchange system packaged inside a dry disposable cassette. E-Gel technology does not required any gel or buffer preparation or gel staining steps. Just load your samples and run.
Live monitoring of DNA migration
E-Gel EX Double Comb gels include SYBR Gold II DNA gel stain with an excitation wavelength in the blue-light spectrum. When used on the E-Gel Power Snap Electrophoresis Device with integrated blue-light trans-illuminator, less-hazardous real-time monitoring of DNA migration is enabled without risk to eyes or possible damage of the sample, unlike UV illumination.
Easy access to the gel
E-Gel EX Double Comb cassettes are designed for convenient opening with a Gel Knife (Cat. No. EI9010), for easy excision of specific bands or transfer of the gel to a membrane for southern blot analysis.
Running and imaging devices
E-Gel EX Double Comb gels require the Invitrogen E-Gel Power Snap Electrophoresis System for gel running and viewing. The system consists of an electrophoresis device and a camera for fast and convenient E-Gel EX Double Comb gel separation and analysis.
Available in a variety of gel formats
E-Gel agarose gels are available in variety of gel percentage, stain, and well formats. The 22-well E-Gel EX Double Comb gels are available in 1% and 2% gel percentages.