Informational:Inquire about OEM or Commercial Supply version of this product here.
Applied Biosystems™

MagMAX™ Viral/Pathogen Binding Solution

Catalog number: A42359
Have Questions?
Applied Biosystems™

MagMAX™ Viral/Pathogen Binding Solution

Catalog number: A42359
Have Questions?
Catalog Number
A42359
Unit Size
550 mL
Price (USD)
Quantity
Catalog NumberUnit SizePrice (USD)Quantity
A42359550 mL
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
This binding buffer is included in the MagMAX Viral/Pathogen Nucleic Acid Isolation kits and MagMAX Microbiome Ultra Nucleic Acid Isolation Kit (Cat. Nos. A42352, A42356, A42357, and A42358). It is made available separately for applications that require more binding buffer than is provided in the kit.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Sample Type
Bacteria, Blood, Buccal Samples, Cells, Fungi, Liquid Samples (e.g. Serum), Virus, Yeast
For Use With (Application)
Next-Generation Sequencing, RT-PCR (Endpoint), Real-Time PCR
High-throughput Compatibility
High-throughput Compatible, Manual Protocols, Automated Protocols
Product Line
MagMAX™
Product Type
Viral/Pathogen Binding Solution
Quantity
550 mL
Shipping Condition
Room Temperature
Target
Viral DNA⁄RNA, Pathogen DNA⁄RNA, Total Nucleic Acids
Unit Size
550 mL

Contents & Storage

Store at room temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text