Informational:Inquire about OEM or Commercial Supply version of this product here.
Applied Biosystems™

MagMAX™ Viral/Pathogen Wash Solution

Catalog number: A42360
Have Questions?
Applied Biosystems™

MagMAX™ Viral/Pathogen Wash Solution

Catalog number: A42360
Have Questions?
Catalog Number
A42360
Unit Size
1000 mL
Price (USD)
Quantity
Catalog NumberUnit SizePrice (USD)Quantity
A423601000 mL
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
This wash solution is included in the MagMAX Viral/Pathogen Nucleic Acid Isolation kits and MagMAX Microbiome Ultra Nucleic Acid Isolation Kit (Cat. Nos. A42352, A42356, A42357, and A42358). It is made available separately for applications that require more wash solution than is provided in the kit.

Important: This wash solution may develop inert white or brown particulates that float in solution. This is not a cause for concern and does not negatively affect performance.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Sample Type
Serum, Viral Samples, Buccal Samples, Blood, Cells, Bacteria, Fungi, Yeast
Capacity
1000 mL
For Use With (Application)
Next-Generation Sequencing, RT-PCR (Endpoint), Real-Time PCR
High-throughput Compatibility
High-throughput Compatible, Manual Protocols, Automated Protocols
Product Line
MagMAX™
Purification Target
Viral DNA⁄RNA, Pathogen DNA⁄RNA, Total Nucleic Acids
Shipping Condition
Room Temperature

Contents & Storage

Store at room temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text