Informational:Inquire about OEM or Commercial Supply version of this product here.
Applied Biosystems™

MagMAX™ Microbiome Lysis Solution

Catalog number: A42361
Have Questions?
Applied Biosystems™

MagMAX™ Microbiome Lysis Solution

Catalog number: A42361
Have Questions?
Catalog Number
A42361
Unit Size
800 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A42361800 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
This lysis buffer is included in the MagMAX Microbiome Ultra Nucleic Acid Isolation Kit (Cat. Nos. A42357, and A42358). It is made available separately for applications that require more lysis solution than is provided in the kit.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Sample Type
Bacteria, Blood, Buccal Samples, Cells, Fungi, Liquid Samples (e.g. Serum), Viral Samples, Yeast
For Use With (Application)
Next-Generation Sequencing, RT-PCR (Endpoint), Real-Time PCR
High-throughput Compatibility
High-throughput Compatible, Manual Protocols, Automated Protocols
Product Line
MagMAX™
Shipping Condition
Room Temperature
Target
Viral DNA⁄RNA, Pathogen DNA⁄RNA, Total Nucleic Acids

Contents & Storage

Store at room temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text