Informational:Inquire about OEM or Commercial Supply version of this product here.
Applied Biosystems™

MagMAX™ Viral/Pathogen Elution Buffer

Catalog number: A42364
Have Questions?
Applied Biosystems™

MagMAX™ Viral/Pathogen Elution Buffer

Catalog number: A42364
Have Questions?
Catalog Number
A42364
Unit Size
100 mL
Price (USD)
Quantity
Catalog NumberUnit SizePrice (USD)Quantity
A42364100 mL
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
This elution buffer is included in the MagMAX Viral/Pathogen Nucleic Acid Isolation kits and MagMAX Microbiome Ultra Nucleic Acid Isolation Kit (Cat. Nos. A42352, A42356, A42357, and A42358). It is made available separately for applications that require more elution buffer than is provided in the kit.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Sample Type
Serum, Viral Samples, Buccal Samples, Blood, Cells, Bacteria, Fungi, Yeast
Capacity
100 mL
For Use With (Application)
Next-Generation Sequencing, RT-PCR (Endpoint), Real-Time PCR
High-throughput Compatibility
High-throughput Compatible, Manual Protocols, Automated Protocols
Product Line
MagMAX™
Purification Target
Viral DNA⁄RNA, Pathogen DNA⁄RNA, Total Nucleic Acids
Quantity
100 mL
Shipping Condition
Room Temperature
Unit Size
100 mL

Contents & Storage

Store at room temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text