The EVOS M7000 Imaging System is a high-performance, fully automated, inverted, multi-channel fluorescence and transmitted light imaging system. It was developed to meet today’s expectations for image quality, user interface interactions, data generation speed and flexibility, and end-user value. The powerful and time-saving acquisition tools with enhanced autofocus algorithms and automated routines for microwell plate assays, time-lapse live cell imaging, area scanning in time lapse and Z-stacks in scan mode, and a superb optomechanical design enable the generation of publication-quality images and data with just a few clicks when and where needed.
This package includes the EVOS M7000 Imaging System, SmartStart orientation, and an extended warranty. The EVOS M7000 Imaging System Extended Warranty Package offers these advantages:
• Outstanding image quality and versatility with a 5-position objective turret, 4-color LED fluorescence and transmitted light channels, and 3.2 MP CMOS color and B/W cameras
• Exceptional usability with fully automated X/Y scanning stage, autofocus, and acquisition routines
• High-speed image acquisition coupled with multi-position well scanning, and Z-stack and tile-stitch options lend power and flexibility to your data generation
• Fully integrated time-lapse live cell imaging using the optional EVOS Onstage Incubator for precise control of temperature, gases for normoxic or hypoxic conditions, and humidity
• Powerful image analysis capabilities for cell segmentation and quantification with the optional Celleste™ Image Analysis Software
• 1-year extended warranty including one planned maintenance visit
Service plans help protect your investment• Maximize system uptime with fast repair turnaround time from manufacturer-trained and certified field service engineers
• Reduce overall repair costs with remote instrument diagnosis and resolution
• Extend the life of your instrument and help keep it running at peak performance with annual maintenance service
Get up and running quickly with SmartStartThe EVOS M7000 Imaging System Extended Warranty Package includes one day of on-site SmartStart orientation carried out by a Field Application Scientist to get you up and running quickly in your lab and reduce downtime. SmartStart is the first step of your education journey and is designed to help you work smarter and with greater efficiency. It includes:
• Basic setup
• Review of hardware and fluidics
• Instrument operation and maintenance
• Software use
• Sample preparation
• Applications
• Data analysis
• Troubleshooting
Content will vary depending on your level of knowledge and experience.
Fully automated imaging systemThe EVOS M7000 system combines and integrates precision components with a unique modern design functionality that enables high-quality automated fluorescence imaging anywhere with unprecedented workflow efficiency. Full automation of the precision X/Y-stage movement; changing of the 4 LED fluorescent light cubes, 5-position objective turret, focus, and exposure; and switching of the dual camera make the EVOS M7000 system an exceptional platform for a variety of demanding imaging applications. Through the intuitive acquisition interface, you can program the EVOS M7000 system to run well-plate scanning, time lapse experiments, and tile-stitch/montage area scans in Z-stack and/or time-lapse modes in your vessel of choice. For live cell imaging over multiple hours or days, the optional EVOS Onstage Incubator, which functions as an environmental chamber on top of the microscope stage, is operated seamlessly from within the instrument interface. The EVOS M7000 system is compatible with all accessories for the predecessor EVOS FL Auto imaging system.
Versatile and highly configurableWith a 5-position objective turret and the ability to simultaneous acquire images in four fluorescence channels plus transmitted light, the optical system can be easily configured to meet your needs. Choose from our broad range of high quality coverslip-corrected and long-working-distance
objectives for use with plastic vessels or glass sides. Available from 1.25X to 100X magnification in achromat, fluorite, or apochromat models, together with over twenty
LED light cubes, the sample imaging options are limitless. Dual monochrome and color high-sensitivity dual CMOS cameras enable seamless imaging of both fluorescent and chromogenically stained samples. Interchangeable
vessel holders accommodate most vessel types and sizes, including slides, multi-well plates, culture flasks, and Petri dishes, and afford precise control and sample alignment by the automated stage. The vessel creation wizard in the user interface enables creation of profiles for custom sample vessels.
Powerful softwareThe EVOS M7000 software presents users with an extensive suite of tools for acquisition, visualization, and analysis. The user interface was designed with a strong emphasis on usability, including an option to view samples in a single field mode (Field View) or in a window that covers a much larger area (Area View). By using a zoom function, the Area View size can be adjusted. A user can explore a large sample area at low magnification, define regions of interest, and then seamlessly transition to the Scan function in Automate Mode and execute an area scan at higher magnification.
The enhanced power of the EVOS M7000 system enables more demanding automated acquisition routines like area scanning in time lapse and Z-stacks in scan mode to be performed. Visualization capabilities include multi-field image viewing gallery, mosaic tiling and tile stitching options, Z-stack visualization, and a video recording and movie maker/editor for time lapse sequences. The EVOS M7000 system is fully compatible with the EVOS Onstage Incubator environmental chamber for control of temperature, gases, and humidity. With the ability generate both hypoxic and normoxic conditions, it is possible to study live cells over extended periods with highly flexible time lapse automation routines.
For users needing more powerful image analysis capabilities, images acquired on the EVOS M7000 system can be analyzed directly using the optional
Celleste Image Analysis Software. Celleste software offers powerful tools for image segmentation and classification that can be applied to a range of cell analysis applications for counting and measuring intensity, area, and shape changes over time, and more.
Smart LED illumination technologyAll EVOS fluorescence imaging systems utilize our proprietary LED light cubes. This light engine outputs remarkable intensity over a short light path and delivers incomparable fluorophore excitation. Each light cube contains a precisely matched set of optical components to optimize the position, evenness, and intensity of the light beam. Digitally controlled LED light sources allow adjustment of illumination levels to the sample and experimental conditions to minimize the risk of photo-toxicity and photo-bleaching in time-lapse studies. Hard-coated filters give sharper edges and significantly higher transmission efficiencies than traditional soft-coated filters.
Easy to use and reliableThe EVOS M7000 Imaging System is powerful, yet offers the simplicity of plug-and-play. Like other EVOS systems, it requires no warm-up or cool-down periods. The LED light source guarantees exceptional stability and durability, so you can turn the unit on and off whenever you need to image a sample. The environmentally safe, mercury-free LED bulbs are rated for >50,000 hours, compared to 300 hours for a typical mercury bulb and 1,500 hours for a metal halide bulb. The long lifetime and low energy consumption translate into significantly lower operating costs compared to instruments with conventional light sources. The EVOS M7000 system is powered by an external Dell PC with an Intel Core i7-8700 processor, 32 GB DDR4 RAM, 512 GB PCIe SSD, and NVIDIA Quadro P1000 with 4 GB discrete video graphics running Windows 10, designed to operate with touchscreen monitor and microscope. Whether acquiring single images or setting up automated routines, the EVOS M7000 system is remarkably easy to use and run. The 1920 x 1080 pixel resolution 23' LCD touch screen monitor enables “finger swipes” to expand or zoom in on images, and moving between microwell plate wells is done by a single touch. Operation is also also fully controllable via mouse.
The EVOS imaging systems are built from the ground up to maximize performance and optimize workflow. You will be astonished at how easy it is to operate and amazed at how extraordinary your images look on-screen.
Explore the entire EVOS line of imaging systems, accessories, and offers ›System and hardware specificationsOptics: infinity‐corrected optical system; RMS‐threaded objectives with 45-mm parfocal distance
For Research Use Only. Not for use in diagnostic procedures.