eBioscience™ Essential Human Treg Phenotyping Kit
eBioscience™ Essential Human Treg Phenotyping Kit
Invitrogen™
eBioscience™ Essential Human Treg Phenotyping Kit
The Invitrogen eBioscience Essential Human Treg Phenotyping Kit is designed for flow cytometry-based identity testing of regulatory T-cells (Tregs). ItRead more
Have Questions?
Catalog number A42925
Price (USD)/ Each
1,446.00
-
Add to cart
Price (USD)/ Each
1,446.00
Add to cart
eBioscience™ Essential Human Treg Phenotyping Kit
Catalog numberA42925
Price (USD)/ Each
1,446.00
-
Add to cart
The Invitrogen eBioscience Essential Human Treg Phenotyping Kit is designed for flow cytometry-based identity testing of regulatory T-cells (Tregs). It leverages high-quality eBioscience antibodies, isotype controls, and fixation/permeabilization reagents (for the intracellular marker FOXP3) and includes a ready-to-use protocol for experimental setup, gating strategy, and analysis.
Benefits include:
Proven performance―antibodies and supporting reagents designed for flow cytometry
Highly referenced antibodies―includes one of the most referenced clones for FOXP3
Turnkey kit and protocol―contains reagents and standardized gating recommendations needed to perform flow cytometry characterization
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence
FormatKit
Quantity100 reactions
Shipping ConditionApproved for shipment at Room Temperature or on Wet or Dry Ice
Product LineeBioscience™
Product TypePhenotyping Kit
Unit SizeEach
Contents & Storage
  • CD4, CD25, CD127, and FOXP3 antibodies, store at 2–8°C
  • FOXP2/TF Staining Buffer set, store at 2–8°C
  • Isotype control, store at 2–8°C