Invitrogen™

Attune™ NxT Maintenance Kit

Catalog number: A43038
Have Questions?
Invitrogen™

Attune™ NxT Maintenance Kit

Catalog number: A43038
Have Questions?
Catalog Number
A43038
Unit Size
1 kit
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A430381 kit
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Attune NxT Maintenance Kit will help you avoid down-time and ensure optimal operation of your Attune NxT Acoustic Focusing Cytometer hardware. This convenient kit includes sufficient supplies to perform all recommended preventative maintenance procedures for 12 months. These procedures are: replacement of the syringe (bi-yearly), SIP tube (yearly), and instrument bottles (yearly), as well as performance of system decontamination (quarterly). The kit includes 1 waste bottle, 1 focusing fluid tank, 12 focusing fluid filters, 2 syringes, 1 SIP tube, and 1 debubble solution.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Block Format
Non-Interchangeable
Product Line
Attune™
Quantity
1 kit
Shipping Condition
Approved for shipment at Room Temperature or on Wet Ice
For Use With (Equipment)
Attune Acoustic Focusing Cytometer
Product Type
Maintenance Kit

Contents & Storage

• 1 waste bottle
• 1 focusing fluid bottle
• 12 focusing fluid filters
• 2 syringes
• 1 SIP
• 1 debubble solution

Store at room temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text