Applied Biosystems™

Pharma KingFisher™ Flex 96 Standard Plates

Catalog number: A43076
Have Questions?
Applied Biosystems™

Pharma KingFisher™ Flex 96 Standard Plates

Catalog number: A43076
Have Questions?
Catalog Number
A43076
Unit Size
48 plates
Price (USD)
Quantity
Catalog NumberUnit SizePrice (USD)Quantity
A4307648 plates
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
These standard plates are for use with the Pharma KingFisher Flex 96 Deep-Well Head Magnetic Particle Processor or MagMAX Express-96 Standard Magnetic Particle Processor in combination with PrepSEQ ResDNA kits and ProteinSEQ kits. The standard configuration differs from the deep-well configuration in that the volume of sample input is from 20–200 μL per well compared to 100–1000 μL per well for the deep-well configuration.

Optimize performance using KingFisher Flex System consumables designed specifically for use with these instruments. KingFisher Flex disposable plastics (tip combs and microplates) are made of polypropylene and are ideal for magnetic particle processing because of their low binding affinity for biomolecules. Their special design enables excellent recovery of magnetic beads.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Barcode
No Barcode
Packaging
48 Plates
Product Line
KingFisher Flex
Color
Clear
Product Type
Microplate
For Use With (Equipment)
KingFisher™ Flex
Material
Polypropylene
Volume (Metric) Well
1000 μL
No. of Wells
96
Quantity
48 plates
Shipping Condition
Room Temperature
Unit Size
48 plates

Contents & Storage

48 standard-well 96-well plates

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text