Informational:TrueCut Cas9 is now available as a CTS-prototype for preclinical and translational research. Learn more »
Applied Biosystems™

PureQuant™ Treg Assay

Catalog number: A43675
Have Questions?
Applied Biosystems™

PureQuant™ Treg Assay

Catalog number: A43675
Have Questions?
Catalog Number
A43675
Unit Size
28 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A4367528 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Applied Biosystems PureQuant assays are a family of assays that accurately identify and quantify specific immune cell types in a mixed population of cells. These assays are TaqMan qPCR-based and measure genomic DNA methylation at specific sites that serve as a unique identifier. The assays include standards and controls and each 96-well plate can accommodate up to seven test samples. Results are provided as a percentage of target cell type out of total input cells using a free analysis tool.

The PureQuant Treg Assay utilizes a specific demethylated site in the transcription factor FOXP3 gene to positively identify regulatory T-cells. This specific site allows for discrimination of regulatory T-cells from activated CD4+ FOXP3+ conventional T-cells (see https://onlinelibrary.wiley.com/doi/full/10.1002/eji.200737594).

Benefits include:
Proven performance―core technology has been used for identification and quantification of immune cell types for over a decade
Easily standardized―qPCR method enables consistent results across multiple users, instruments, and sites
Facilitates regulatory filings―established performance specifications following ICHQ2(R1) guidelines
Scheduling flexibility―assay can be performed using fresh or frozen cells/gDNA

The assays include all reagents, standards, and controls required to run the assay. Specific components include:
Calibrator: corrects for variation in bisulfite conversion efficiency. Plasmid-based calibrator harbors the genomic target regions in equimolar ratios and includes GAPDH. Subjected to bisulfite conversion followed by copy number estimation via qPCR standard curve. Allows estimation of assay-specific bisulfite efficiency factor: the ratio between TpG/GAPDH copy numbers measured from the same plasmid.
Reference: checks assay performance. Consists of genomic DNA extracted from human blood of pooled healthy donors. The reference values represent the expected range of the target cell type. For example, CD8+ cells are generally more abundant than Treg or Th17 cells.
Standards: determine the unknown copy number. Consist of six dilutions spanning 4-log linear dynamic range (30-31250 copies). Absolute copy numbers can reliably be determined using the slope and intercept of the respective standard curves.

This product is manufactured at a site with an ISO9001-certified quality management system.

Specifications

Detection Method
Primer-Probe Detection
For Use With (Application)
Cells, DNA
No. of Tests
28
Shipping Condition
Approved for shipment at Room Temperature or on Wet or Dry Ice

Contents & Storage

  • Dynabeads SILANE Genomic DNA Kit, store at 2°C to 8°CC
  • 1 x 2 mL PureQuant qPCR Master Mix, store at 2°C to 8°CC
  • 4 x ABS
  • THFA, store at room temperature
  • PureQuant Treg Module, store at -15 to -25°C (FOXP3 & GAPDH TaqMan primers and probes, Reference DNA, Lambda DNA, Standard DNA)

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text