Thermo Scientific™

Dual QuantStudio System Integration Kit for the Orbitor™ RS2 Microplate Mover and QuantStudio™ Real-time PCR Systems

Catalog number: A43779
Have Questions?
Thermo Scientific™

Dual QuantStudio System Integration Kit for the Orbitor™ RS2 Microplate Mover and QuantStudio™ Real-time PCR Systems

Catalog number: A43779
Have Questions?
Catalog Number
A43779
Unit Size
1 kit
Price (USD)
Catalog NumberUnit SizePrice (USD)
A437791 kitRequest A Quote
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
This Dual QuantStudio System Integration Kit is designed for use with the Orbitor RS2 Microplate Mover and identical QuantStudio Systems, enabling real-time PCR in automated environments for increased sample throughput.

This integration kit includes a location bracket, splitter, and Thermo Scientific Momentum Workflow Software for the second QuantStudio integration.

Couple the Dual QuantStudio Integration Kit with the Orbitor RS2 Microplate Mover for QuantStudio Systems and one of the two plate storage and handling options (Sequential Access or Random Access ) to complete your automation bundle.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment)
QuantStudio 7 Pro, QuantStudio 7 Flex, or QuantStudio 12 K Flex Real-Time PCR Systems and The Orbitor RS2 Microplate Mover
Product Type
Integration Kit
For Use With (Application)
Real Time PCR (qPCR)
Shipping Condition
Room Temperature
Quantity
1 kit

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text