Thermo Scientific™

Orbitor™ RS2 Microplate Mover for QuantStudio™ Real-time PCR Systems

Catalog number: A43780
Have Questions?
Thermo Scientific™

Orbitor™ RS2 Microplate Mover for QuantStudio™ Real-time PCR Systems

Catalog number: A43780
Have Questions?
Catalog Number
A43780
Unit Size
1 mover
Price (USD)
Catalog NumberUnit SizePrice (USD)
A437801 moverRequest A Quote
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
This Orbitor RS2 Microplate Mover is designed for use with the QuantStudio 7 Pro, QuantStudio 7 Flex, and QuantStudio 12K Flex real-time PCR systems, enabling real-time PCR in automated environments for increased sample throughput.

Extensive vertical reach allows the robot to access virtually any microplate instrument location, including stacked instruments, and its telescoping arm can reach a variety of distances. The ingenious arm travels right through the base, resulting in rapid and efficient movements. The Orbitor RS2 Microplate Mover has an integrated barcode scanner for greater flexibility and a full 360°workspace with unlimited base rotation that eliminates the need for hard stops and reverse movements.

Advanced closed-loop motion control provides collision detection and position monitoring, allowing the mover to quickly resume without the need to re-home. With a wide range of grip-control functionality, this mover combines performance with innovation. From variable force control to plate detection, the feature-set is designed to impress. The result is remarkable speed, precise motion, and consistent, accurate plate placement.

The Orbitor RS2 Microplate Mover is powered by Thermo Scientific Momentum Workflow Scheduling Software, combining power with ease of use.

Couple this Orbitor RS2 Microplate Mover with the applicable pre-configured plate storage and handling option for the QuantStudio systems to complete your automation bundle. Two plate storage and handling options are available: sequential access stacks (40 plates/3 stacks) and random access hotels (24 plates/3 hotels).

For integration of two QuantStudio systems with the Orbitor RS2 mover, the Dual QuantStudio System Integration Kit is required in addition to the automation bundle.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment)
QuantStudio 7 Pro, QuantStudio 7 Flex, or QuantStudio 12 K Flex Real-Time PCR Systems
Product Type
Microplate Mover
For Use With (Application)
Real Time PCR (qPCR)
Shipping Condition
Room Temperature
Quantity
1 Mover

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text