Thermo Scientific™

Sequential Access Storage Option for the Orbitor™ RS2 Microplate Mover for QuantStudio™ Real-time PCR Systems

Catalog number: A43781
Have Questions?
Thermo Scientific™

Sequential Access Storage Option for the Orbitor™ RS2 Microplate Mover for QuantStudio™ Real-time PCR Systems

Catalog number: A43781
Have Questions?
Catalog Number
A43781
Unit Size
1 each
Price (USD)
Catalog NumberUnit SizePrice (USD)
A437811 eachRequest A Quote
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
This Sequential Access Storage Option is designed for use with the Orbitor RS2 Microplate Mover and a QuantStudio real-time PCR system, enabling real-time PCR in automated environments for increased sample throughput.

This storage option holds up to 20 plates/3 stacks (96-well) and up to 40 plates/3 stacks (384-well). These stacks provide the option for the Orbitor RS2 mover to “run until empty”, which allows for additional plates to be added while the Orbitor RS2 mover is running. This option is ideal for 0.1-mL and 0.2-mL 96-well plates and 384-well plates.

Couple this Sequential Access Storage Option with the Orbitor RS2 Microplate Mover for QuantStudio Systems to complete your automation bundle.

For integration of two QuantStudio systems with the Orbitor RS2 mover, the Dual QuantStudio System Integration Kit is required in addition to the automation bundle.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment)
QuantStudio 7 Pro, QuantStudio 7 Flex, or QuantStudio 12 K Flex Real-Time PCR Systems and The Orbitor RS2 Microplate Mover
For Use With (Application)
Real Time PCR (qPCR)
Shipping Condition
Room Temperature
Description
Storage Option
Quantity
1 Ea.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text